Skip to Content
Merck
All Photos(1)

Key Documents

63005BC

Millipore

Supplement V, SimPlate®

BioControl, For use with SimPlate® Yeast & Mold Color Indicator

Synonym(s):

Fluorogenic Indicator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.85

Pricing and availability is not currently available.

material

plastic vial (containing dehydrated culture media)

form

liquid

shelf life

6‑36 mo.

packaging

bottle of 1 20 mL

manufacturer/tradename

BioControl Systems 63005-200

storage condition

protect from light

application(s)

food and beverages

compatibility

for use with SimPlate (66007BC)

storage temp.

2-8°C

suitability

molds
yeasts

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
63004BC63012BC63010BC
manufacturer/tradename

BioControl Systems 63005-200

manufacturer/tradename

BioControl Systems 63004-200

manufacturer/tradename

BioControl Systems 63012-100

manufacturer/tradename

BioControl Systems 63010-200

packaging

bottle of 1 20 mL

packaging

bottle of 1 20 mL

packaging

bottle of 1 20 mL

packaging

bottle of 1 40 mL

form

liquid

form

liquid

form

liquid

form

liquid

shelf life

6‑36 mo.

shelf life

6‑36 mo.

shelf life

6‑15 mo.

shelf life

6‑36 mo.

storage condition

protect from light

storage condition

protect from light

storage condition

protect from light

storage condition

protect from light

General description

Supplement V is an additional reagent for use in the SimPlate® yeast & mold. SimPlate® Binary Detection Technology combines proprietary media and a patented plating device to provide fast and accurate quantitative results for key groups of indicator organisms. Defined substrates incorporated into the media are utilized to detect multiple enzymes and redox reactions associated with the target organisms resulting in chromogenic or fluorescent changes to the media, providing easy-to-read results. Supplement V is added to convert color indicator to fluorogenic media when visualized under UV light.

Application

Supplement V is added to SimPlate® trays to convert color indicator to fluorogenic media for the detection of yeast and molds in sample with high vitamin C content such as undiluted juice or dry pet food without the need for optimization of reaction and cycle conditions.

Components

1 bottle, approx. 20 mL (adequate for 200 tests)

Other Notes

For use with SimPlate Yeast & Mold CI (66007BC)

Legal Information

SimPlate is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Customers Also Viewed

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service