63004BC
Supplement M, SimPlate®
BioControl, For use with SimPlate Yeast & Mold Color Indicator
Synonym(s):
Color Indicator
About This Item
Recommended Products
material
plastic vial (containing dehydrated culture media)
form
liquid
shelf life
6‑36 mo.
packaging
bottle of 1 20 mL
manufacturer/tradename
BioControl Systems 63004-200
storage condition
protect from light
application(s)
food and beverages
compatibility
for use with SimPlate (66007BC)
storage temp.
2-8°C
suitability
molds
yeasts
Related Categories
General description
Application
Components
Other Notes
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service