Skip to Content
Merck
All Photos(1)

Key Documents

55952

Supelco

Fusaric acid

for HPLC derivatization, ≥99.0% (HPLC)

Synonym(s):

5-Butylpicolinic acid, 5-Butylpyridine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO2
CAS Number:
Molecular Weight:
179.22
Beilstein:
125804
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.22

grade

for HPLC derivatization

Quality Level

Assay

≥99.0% (HPLC)

technique(s)

HPLC: suitable

SMILES string

CCCCc1ccc(nc1)C(O)=O

InChI

1S/C10H13NO2/c1-2-3-4-8-5-6-9(10(12)13)11-7-8/h5-7H,2-4H2,1H3,(H,12,13)

InChI key

DGMPVYSXXIOGJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fusaric acid is a novel proton-affinitive derivatizing agent, having an ionization moiety and a hydrophobic moiety. It is commonly used for the derivatization of alcohols and phenols, by liquid chromatography coupled with electrospray ionization tandem mass spectrometry (LC/ESI-MS/MS).

Application

Fusaric acid may be used as a derivatizing reagent for the quantification of hydroxysteroids and dehydroepiandrosterone (DHEA) and sulfated DHEA in biological samples using liquid chromatography electrospray-ionization-tandem mass spectrometry (LC/ESI-MS/MS) technique.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical derivatization to enhance ionization of anabolic steroids in LC-MS for doping-control analysis
I. Athanasiadou et all.
TrAC, Trends in Analytical Chemistry, 42, 137-156 (2013)
Kouwa Yamashita et al.
Journal of the American Society for Mass Spectrometry, 21(2), 249-253 (2009-11-17)
A highly sensitive derivatization method for liquid chromatography (LC)-electrospray ionization (ESI) tandem mass spectrometry of dehydroepiandrosterone (DHEA), testosterone (T), pregnenolone (P5), and 17alpha-OH-pregnenolone (17-OHP5) was developed based on the use of fusaric acid as a reagent. DHEA, P5, and 17-OHP5
Sevcan Mamur et al.
Drug and chemical toxicology, 43(2), 149-157 (2018-09-12)
Fusaric acid (FA) is produced by several Fusarium species and is commonly found in grains. This investigation was performed to evaluate the cytotoxic and genotoxic effects of FA either in human cervix carcinoma (HeLa) cell line using 3-(4,5-dimethylthiazolyl-2)-2,5 diphenyltetrazolium bromide
T Santa
Drug discoveries & therapeutics, 7(1), 9-17 (2013-03-26)
Liquid chromatography coupled with electrospray ionization tandem mass spectrometry (LC/ESI-MS/MS) has been frequently utilized for the sensitive and selective determination of the trace level compounds in biological samples. In LC/ESI-MS/MS, chemical derivatization is sometimes used to enhance the detection sensitivity
Yasuhiro Shibata et al.
The Journal of steroid biochemistry and molecular biology, 145, 193-199 (2014-05-06)
A reliable and sensitive method for analyzing steroids using liquid chromatography tandem mass spectrometry (LC-MS/MS) is required for research concerning dehydroepiandrosterone (DHEA), which plays a central role in steroid hormone biosynthesis and metabolism. Furthermore, after the first proposal of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service