Skip to Content
Merck
All Photos(1)

Key Documents

Y0000669

Salicin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

D-(−)-Salicin, 2-(Hydroxymethyl)phenyl-β-D-glucopyranoside, Salicoside, Salicyl alcohol glucoside, Saligenin β-D-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O7
CAS Number:
Molecular Weight:
286.28
Beilstein:
89593
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

salicin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](Oc2ccccc2CO)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C13H18O7/c14-5-7-3-1-2-4-8(7)19-13-12(18)11(17)10(16)9(6-15)20-13/h1-4,9-18H,5-6H2/t9-,10-,11+,12-,13-/m1/s1

InChI key

NGFMICBWJRZIBI-UJPOAAIJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Salicin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanna Oltean et al.
The Cochrane database of systematic reviews, 12(12), CD004504-CD004504 (2014-12-24)
Low-back pain (LBP) is a common condition and imposes a substantial economic burden upon people living in industrialized societies. A large proportion of people with chronic LBP use complementary and alternative medicine (CAM), visit CAM practitioners, or both. Several herbal
Ken Keefover-Ring et al.
PloS one, 9(10), e107189-e107189 (2014-10-10)
Salicinoids are well-known defense compounds in salicaceous trees and careful screening at the population level is warranted to fully understand their diversity and function. European aspen, Populus tremula, is a foundation species in Eurasia and highly polymorphic in Sweden. We
Mahla Mirali et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 969, 149-161 (2014-09-01)
A systematic set of optimization experiments was conducted to design an efficient extraction and analysis protocol for screening six different sub-classes of phenolic compounds in the seed coat of various lentil (Lens culinaris Medik.) genotypes. Different compounds from anthocyanidins, flavan-3-ols
Véronique Gilard et al.
Journal of pharmaceutical and biomedical analysis, 102, 476-493 (2014-12-03)
One hundred and fifty dietary supplements (DS) marketed to increase sexual performance were analyzed. All these formulations were claimed to contain only natural compounds, plant extracts and/or vitamins. (1)H NMR spectroscopy was used for detecting the presence of adulterants and
Min Jung Kim et al.
Biochemical and biophysical research communications, 456(2), 586-590 (2014-12-10)
Taste-taste interactions often showed in human psychophysical studies. Considering that each tastant in foodstuffs individually stimulates its responsible gustatory systems to elicit relevant taste modalities, taste-taste interaction should be performed in taste receptor cell-based assay. While umami substances have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service