Skip to Content
Merck
All Photos(1)

Key Documents

51381

Supelco

(±)-β-Citronellol

analytical standard

Synonym(s):

3,7-Dimethyl-6-octen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CH2OH
CAS Number:
Molecular Weight:
156.27
Beilstein:
1721507
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

5.4 (vs air)

vapor pressure

~0.02 mmHg ( 25 °C)

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.456 (lit.)

bp

222 °C (lit.)

density

0.857 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC(CCO)CC\C=C(\C)C

InChI

1S/C10H20O/c1-9(2)5-4-6-10(3)7-8-11/h5,10-11H,4,6-8H2,1-3H3

InChI key

QMVPMAAFGQKVCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certan Vial

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Lapczynski et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46 Suppl 11, S103-S109 (2008-07-22)
A toxicologic and dermatologic review of dl-citronellol when used as a fragrance ingredient is presented.
Renan G Brito et al.
Journal of natural medicines, 66(4), 637-644 (2012-02-22)
We describe the antinociceptive and anti-inflammatory properties of citronellol (CT) in rodents. CT, a monoterpene alcohol, is a naturally occurring monoterpene compound prevalent in essential oils of various aromatic plant species, such as Cymbopogon citratus. In mice, when evaluated against
Michiko Katsukawa et al.
Bioscience, biotechnology, and biochemistry, 75(5), 1010-1012 (2011-05-21)
We evaluated the effects of rose oil on the peroxisome proliferator-activated receptor (PPAR) and cyclooxygenase-2 (COX-2). Citronellol and geraniol, the major components of rose oil, activated PPARα and γ, and suppressed LPS-induced COX-2 expression in cell culture assays, although the
Yoshikazu Kitano et al.
Biofouling, 27(2), 201-205 (2011-02-01)
Twenty novel simple alkyl isocyanides derived from citronellol were synthesized and evaluated for their antifouling activity and toxicity against cypris larvae of the barnacle, Balanus amphitrite. The anti-barnacle activity of the synthesized isocyanides was in the EC(50) range of 0.08-1.49
Wantida Chaiyana et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(8-9), 836-839 (2012-04-19)
Inhibition of cholinesterase has attracted much attention recently because of its potential for the treatment of Alzheimer's disease. In this work, the anticholinesterase activities of plant oils were investigated using Ellman's colorimetric method. The results indicate that essential oils obtained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service