Skip to Content
Merck
All Photos(1)

Key Documents

46426

Supelco

S-Hydroprene

PESTANAL®, analytical standard

Synonym(s):

Ethyl 3,7,11-trimethyl-2,4-dodecadienoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H30O2
CAS Number:
Molecular Weight:
266.42
Beilstein:
5255711
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOC(=O)\C=C(C)\C=C\C[C@@H](C)CCCC(C)C

InChI

1S/C17H30O2/c1-6-19-17(18)13-16(5)12-8-11-15(4)10-7-9-14(2)3/h8,12-15H,6-7,9-11H2,1-5H3/b12-8+,16-13+/t15-/m0/s1

InChI key

FYQGBXGJFWXIPP-OJROSNHMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Said A M Mazyad et al.
Journal of the Egyptian Society of Parasitology, 36(3), 1101-1114 (2006-12-13)
Two insect growth regulators, Methorprene and Hydroprene were evaluated as growth hormones on the common house dust mite; Dermatophagoides farinae. The adults and immature stages were treated with 4 concentrations of each regulator as contact and as diet. The data
Yuichiro Suzuki et al.
Development (Cambridge, England), 135(3), 569-577 (2008-01-04)
The evolution of complete metamorphosis in insects is a key innovation that has led to the successful diversification of holometabolous insects, yet the origin of the pupa remains an enigma. Here, we analyzed the expression of the pupal specifier gene
Chloe Hawkings et al.
PeerJ, 6, e4875-e4875 (2018-06-06)
Vitellogenin has been proposed to regulate division of labor and social organization in social insects. The red imported fire ant (Solenopsis invicta) harbors four distinct, adjacent vitellogenin genes (Vg1, Vg2, Vg3, and Vg4). Contrary to honey bees that have a
B L Reid et al.
Journal of economic entomology, 87(6), 1537-1546 (1994-12-01)
The influence of sterilization by hydroprene on population dynamics in the German cockroach, Blattella germanica (L.), was studied in the laboratory where more detailed and accurate assessments could be achieved than would be possible under typical field situations. The gradual
Frank H Arthur et al.
Pest management science, 65(7), 791-797 (2009-04-11)
Pyriproxyfen and hydroprene are insect growth regulators (IGRs) that have been evaluated to control insect pests of field crops, but there are limited reports of efficacy against stored-product insects. A laboratory study was conducted to determine residual efficacy of pyriproxyfen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service