Skip to Content
Merck
All Photos(2)

Key Documents

45795

Supelco

1-Methylnaphthalene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH3
CAS Number:
Molecular Weight:
142.20
Beilstein:
506793
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98%

autoignition temp.

984 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.615 (lit.)

bp

240-243 °C (lit.)

mp

−22 °C (lit.)

density

1.001 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Cc1cccc2ccccc12

InChI

1S/C11H10/c1-9-5-4-7-10-6-2-3-8-11(9)10/h2-8H,1H3

InChI key

QPUYECUOLPXSFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Methylnaphthalene defines the lower (zero) reference point of cetane number, a measure of diesel fuel ignition quality, as it has a long ignition delay (poor ignition qualities)
The 1-Methylnaphthalene may find the following uses:
  • Study of surface and electronic features of fluorinated TiO2 and their influence on the photocatalytic degradation of 1-methylnaphthalene
  • Thermal decomposition of 1-Methylnaphthalene in a batch reactor
  • Catalytic conversion of tar from hot coke oven gas using 1-methylnaphthalene as a tar model compound
  • Hydrocracking of 1-methylnaphthalene to benzene, toluene, and xylene using high metal–acid balance and selective hydrogenation activity catalysts

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas purge microsyringe extraction for quantitative direct gas chromatographic-mass spectrometric analysis of volatile and semivolatile chemicals.
Yang CP, et al.
Journal of Chromatography A, 1218(12), 1549-1555 (2011)
Analysis of aromatic compounds in gasoline with flow-switching comprehensive two-dimensional gas chromatography.
Micyus NJ, et al.
Journal of Chromatography A, 1086(1-2), 115-121 (2005)
Development and validation of a method for air-quality and nuisance odors monitoring of volatile organic compounds using multi-sorbent adsorption and gas chromatography/mass spectrometry thermal desorption system.
Ribes A, et al.
Journal of Chromatography A, 1140(1-2), 44-55 (2007)
Comparative study of solvent extraction and thermal desorption methods for determining a wide range of volatile organic compounds in ambient air.
Ramirez N, et al.
Talanta, 82(2), 719-727 (2010)
Evaluation of an in-injection port thermal desorption-gas chromatography/mass spectrometry method for analysis of non-polar organic compounds in ambient aerosol samples.
Ho SSH, et al.
Journal of Chromatography A, 1200(2), 217-227 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service