Skip to Content
Merck
All Photos(1)

Key Documents

45671

Supelco

Tebuthiuron

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16N4OS
CAS Number:
Molecular Weight:
228.31
Beilstein:
527479
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(=O)N(C)c1nnc(s1)C(C)(C)C

InChI

1S/C9H16N4OS/c1-9(2,3)6-11-12-8(15-6)13(5)7(14)10-4/h1-5H3,(H,10,14)

InChI key

HBPDKDSFLXWOAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tebuthiuron is a thiadiazolylurea derivative and a root active, soil applied herbicide, effectively used for woody plant control.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tebuthiuron may be used as an analytical reference standard for the quantification of the analyte in environmental samples using different chromatography techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Milena Devechi de Almeida et al.
Aquatic toxicology (Amsterdam, Netherlands), 194, 86-93 (2017-11-24)
Tebuthiuron is a phenylurea herbicide widely used in agriculture that can reach the aquatic environments, possibly posing negative effects to the aquatic biota. Phenylurea herbicides, such as diuron, are known to cause estrogenic and anti-androgenic effects in fish, but no
Woody Plants and Woody Plant Management: Ecology: Safety, and Environmental ImPatt (2001)
Determination of sugar cane herbicides in soil and soil treated with sugar cane vinasse by solid-phase extraction and HPLC-UV
Lourencetti C, et al.
Talanta, 77(2), 701-709 (2008)
Ming Su et al.
Journal of the science of food and agriculture, 98(13), 4845-4853 (2018-03-27)
Some trace amounts of urea herbicide residues can be transferred to humans via the food chain, thereby being potentially harmful to human health. The development of a robust analytical methodology for effective sample preparation and simultaneous determination of herbicide residues
Marie C Thomas et al.
Scientific reports, 10(1), 19592-19592 (2020-11-13)
Conventional photosystem II (PSII) herbicides applied in agriculture can pose significant environmental risks to aquatic environments. In response to the frequent detection of these herbicides in the Great Barrier Reef (GBR) catchment area, transitions towards 'alternative' herbicides are now widely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service