Skip to Content
Merck
All Photos(1)

Key Documents

45637

Supelco

Propachlor

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-N-isopropylacetanilide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14ClNO
CAS Number:
Molecular Weight:
211.69
Beilstein:
2103903
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)N(C(=O)CCl)c1ccccc1

InChI

1S/C11H14ClNO/c1-9(2)13(11(14)8-12)10-6-4-3-5-7-10/h3-7,9H,8H2,1-2H3

InChI key

MFOUDYKPLGXPGO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propachlor is a herbicide, widely used in oil seeds and crop planting.

Application

Propachlor may be used as an analytical reference standard for the quantification of the analyte in industrial wastewater samples and complex matrices using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquid-liquid extraction for sample preparation prior to gas chromatography and gas chromatography-mass spectrometry determination of herbicide and pesticide compounds
Mahara.MB, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 58(1), 31-38 (1998)
Multiresidue analysis of 240 pesticides in apple and lettuce by QuEChERS sample preparation and HPLC-MS/MS analysis
Wang.L, et al.
The Korean Journal of Pesticide Science, 46(5), 424-429 (2008)
Sheng Tai et al.
Molecular cancer therapeutics, 11(6), 1320-1331 (2012-04-12)
PI3K/AKT/mTOR pathway plays a key role in the tumorigenesis of many human cancers including prostate cancer. However, inhibitors of this pathway, such as Rad001, have not shown therapeutic efficacy as a single agent. Through a high-throughput screen of 5,000 widely
A R Loch et al.
Environmental science & technology, 36(19), 4065-4073 (2002-10-17)
Reactions of bisulfide and polysulfides with alachlor, propachlor, and metolachlor were examined in aqueous solution to investigate the role reduced sulfur species could play in effecting abiotic transformations of chloroacetanilide herbicides. Experiments at 25 degrees C demonstrated that reactions were
M B Genter et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 47(6), 1051-1057 (2009-05-09)
Alachlor and butachlor are chloracetanilide herbicides that induce olfactory tumors in rats, whereas propachlor does not. The mechanism by which alachlor induces tumors is distinct from many other nasal carcinogens, in that alachlor induces a gradual de-differentiation of the olfactory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service