Skip to Content
Merck
All Photos(2)

Key Documents

45322

Supelco

Aminocarb

PESTANAL®, analytical standard

Synonym(s):

(4-Dimethylamino-3-methylphenyl) N-methylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N2O2
CAS Number:
Molecular Weight:
208.26
Beilstein:
2808681
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(=O)Oc1ccc(N(C)C)c(C)c1

InChI

1S/C11H16N2O2/c1-8-7-9(15-11(14)12-2)5-6-10(8)13(3)4/h5-7H,1-4H3,(H,12,14)

InChI key

IMIDOCRTMDIQIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K M Sundaram et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 20(2), 187-200 (1985-04-01)
Three aminocarb formulations, water-based Matacil 180F, oil-based Matacil 180F and oil-based Matacil 180D, were applied separately to the surface of a stream at 2.40 g a.i./min for 5 minutes. The highest aminocarb concentration in the top 1 cm of water
D J Ecobichon
Biomedical and environmental sciences : BES, 3(2), 217-239 (1990-06-01)
The management of insect epidemics in large tracts of forest is difficult given the climatic conditions encountered, the topography of the forested land, the nature of the forest, the types of chemical and/or biological insecticides registered for use, and the
Fenitrothion and aminocarb residues in water and balsam fir foliage following spruce budworm spraying programs in Quebec, 1979 to 1982.
R Morin et al.
Bulletin of environmental contamination and toxicology, 36(4), 622-628 (1986-04-01)
A Sundaram et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 20(2), 167-186 (1985-04-01)
The influence of formulation properties on spray droplet spectra and soil residues was studied in conifer forests in New Brunswick following aerial application of two oil-based aminocarb formulations of variable viscosities and volatilities. For a given volume rate of application
M Fournier et al.
Journal of toxicology and environmental health, 25(1), 103-118 (1988-01-01)
Interaction of two potential immunosuppressive factors, sublethal pesticide exposure and viral inhibition of lymphocyte mitogenesis, was examined in mixed lymphocyte reaction (MLR). Inbred (C57Bl/6 x A/J)F mice, semisusceptible to mouse hepatitis virus 3 (MHV3) infection were exposed to selected pesticides

Protocols

-methylcarbamate 10 μg/mL; Diuron; Propham; Siduron; Methiocarb, analytical standard; Linuron 10 μg/mL; Swep 10 μg/mL; Chlorpropham 10 μg/mL; Barban; Neburon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service