Skip to Content
Merck
All Photos(1)

Key Documents

43648

Supelco

Parathion-methyl

certified reference material, TraceCERT®

Synonym(s):

O,O-Dimethyl O-(4-nitrophenyl) phosphorothioate, Methyl parathion

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10NO5PS
CAS Number:
Molecular Weight:
263.21
Beilstein:
8905814
MDL number:
UNSPSC Code:
41116107

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

SMILES string

COP(=S)(OC)Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C8H10NO5PS/c1-12-15(16,13-2)14-8-5-3-7(4-6-8)9(10)11/h3-6H,1-2H3

InChI key

RLBIQVVOMOPOHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 . This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Parathion methyl is a non-systemic organophosphate insecticide and acaricide. It acts by inhibiting the activity of the acetylcholinesterase (AChE) enzyme, thereby blocking hydrolysis of acetylcholine and leading to hyperexcitation. It is used to control chewing and sucking insects in cereals, fruit, citrus, vines, vegetables, ornamentals, cotton, and field crops.
According to the Commission Implementing Regulation (EU) No 2003/166/EC of 10th March 2003, parathion methyl is not approved for use in the European Union (EU) as a plant protection agent, following the Regulation (EC) No. 1107/2009. But maximum residue levels of 0.01 mg/kg to 5 mg/kg are allowed for the presence of parathion methyl (sum of parathion-methyl and paraoxon-methyl expressed as parathion-methyl) in products of plant and animal origin, according to the EU Reg. No 899/2012.
Parathion methyl has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Parathion methyl CRM may also be used as given below:
  • Development of a fluorescence sensor based on near-infrared CuInS2 quantum dots (QDs) and organophosphorus hydrolase (OPH) to detect parathion methyl in tap water, river water, rice, and banana samples
  • Evaluate polyaniline (PANI)-modified zeolite NaY as a sorbent for the extraction of multiclass pesticides from water, soil, honey, fruit, and vegetable samples and their quantification by high-performance liquid chromatography (HPLC) coupled with photodiode array (PDA) detection
  • Determination of 36 pesticides in soil samples using modified QuEChERS method for extraction and dispersive-solid phase extraction (d-SPE) for clean-up, followed by quantification with liquid chromatography with tandem quadrupole linear ion trap mass spectrometry (LC-MS/MS)
  • Multi-residue analysis of 160 multiclass pesticides using ethyl acetate (EtAC)/n. hexane mixture for extraction from chamomile, thyme, and marjoram samples and their analysis by gas chromatography-tandem mass spectrometry (GC-MS/MS)
  • Optimization of a gas chromatography-flame photometry detector (GC-FPD) based method for the determination of 44 organophosphorous pesticide (OPP) residues in Pogostemon cablin and related product samples
  • Simultaneous residue analysis of 379 pesticides in human serum samples using QuEChERS extraction method and liquid chromatography-tandem mass spectrometry (LC-MS/MS)

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3 - STOT RE 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - open cup

Flash Point(C)

46 °C - open cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service