Skip to Content
Merck
All Photos(1)

Key Documents

43029

Supelco

1,3-Diphenylguanidine

analytical standard (for titrimetry)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5NH)2C=NH
CAS Number:
Molecular Weight:
211.26
Beilstein:
1875653
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
39032512
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard (for titrimetry)

Quality Level

Assay

≥99.5% (HPLC)

technique(s)

titration: suitable

mp

146-148 °C (lit.)
149-150 °C

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

N=C(Nc1ccccc1)Nc2ccccc2

InChI

1S/C13H13N3/c14-13(15-11-7-3-1-4-8-11)16-12-9-5-2-6-10-12/h1-10H,(H3,14,15,16)

InChI key

OWRCNXZUPFZXOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Diphenylguanidine volumetric standard is manufactured under stringent conditions and measured with the highest possible precision.

Application

It finds its use as an analytical standard for the standardization of volumetric solutions in titrimetric analyses.

Features and Benefits

  • Available as a solid in a secure glass bottle to ensure its stability for the entire shelf life until opened.
  • High-quality offering accurate titer determinations
  • Accompanied by a certificate of analysis (CoA)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

338.0 °F - closed cup

Flash Point(C)

170 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solubility determination and thermodynamic dissolution functions of 1, 3-diphenylguanidine in nine organic solvents at evaluated temperatures
Xu R, et al.
The Journal of Chemical Thermodynamics, 99, 86-95 (2016)
Quantification of organic contaminants in urban stormwater by isotope dilution and liquid chromatography-tandem mass spectrometry
Hou F, et al.
Analytical and Bioanalytical Chemistry, 411(29), 7791-7806 (2019)
Allergy to rubber additives in orthopedic braces.
S T Bolhaar et al.
Contact dermatitis, 42(5), 288-289 (2000-05-02)
Mingxiang Lin et al.
Journal of pharmaceutical and biomedical analysis, 45(5), 747-755 (2007-10-12)
During the content uniformity test of a drug product (tablet formulation), an unknown peak was observed in the HPLC chromatograms. Upon further investigation, it was determined that the unknown peak was originated from an external source and, therefore, the drug
H B Koëter et al.
Toxicology, 71(1-2), 173-179 (1992-01-01)
An oral testicular toxicity and male fertility study was carried out in CD-1 mice with 1,3-diphenylguanidine (99.9% purity). 1,3-Diphenylguanidine was administered to male mice by daily gavage at dose levels of 0, 0.06, 0.25, 1, 4 and 16 mg/kg body

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service