Skip to Content
Merck
All Photos(2)

Key Documents

207713

Sigma-Aldrich

1,3-Di-o-tolylguanidine

99%

Synonym(s):

DTG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3C6H4NH)2C(=NH)
CAS Number:
Molecular Weight:
239.32
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

176-178 °C (lit.)

solubility

water: soluble 70 mg/mL at 20 °C

functional group

amine

SMILES string

Cc1ccccc1NC(=N)Nc2ccccc2C

InChI

1S/C15H17N3/c1-11-7-3-5-9-13(11)17-15(16)18-14-10-6-4-8-12(14)2/h3-10H,1-2H3,(H3,16,17,18)

InChI key

OPNUROKCUBTKLF-UHFFFAOYSA-N

Gene Information

human ... EBP(10682)

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Di-o-tolylguanidine is a selective σ-receptor agonist. 1,3-Di-o-tolylguanidine is σ site ligand and its interaction with dopamine drugs were studied in Wistar rats and Albino Swiss mice.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melissa Wares et al.
Antimicrobial agents and chemotherapy, 59(4), 1942-1949 (2015-01-15)
Drug resistance represents a key aspect of human immunodeficiency virus (HIV) treatment failure. It is important to develop nonhuman primate models for studying issues of drug resistance and the persistence and transmission of drug-resistant viruses. However, relatively little work has
Zur Mira Azizah Nor Haiza Lah et al.
Journal of pharmaceutical and biomedical analysis, 174, 608-617 (2019-07-03)
A facile electrochemical sandwich immunosensor for the detection of a breast cancer biomarker, the human epidermal growth factor receptor 2 (HER2), was designed, using lead sulfide quantum dots-conjugated secondary HER2 antibody (Ab2-PbS QDs) as a label. Using Ab2-PbS QDs in
Mona Ebadi et al.
Scientific reports, 10(1), 21521-21521 (2020-12-11)
The use of nanocarriers composed of polyethylene glycol- and polyvinyl alcohol-coated vesicles encapsulating active molecules in place of conventional chemotherapy drugs can reduce many of the chemotherapy-associated challenges because of the increased drug concentration at the diseased area in the
Moya A Hay et al.
Chemical science, 10(25), 6354-6361 (2019-07-26)
The magnetic properties of 3d monometallic complexes can be tuned through geometric control, owing to their synthetic accessibility and relative structural simplicity. Monodentate ligands offer great potential for fine-tuning the coordination environment to engineer both the axial and rhombic zero-field
S A Mousavi et al.
Ultrasonics sonochemistry, 46, 26-35 (2018-05-10)
A nanostructured cationic zinc nitrate complex with a formula of [ZnLNO3]NO3 (where L = (N2E,N2'E)-N1,N1'-(ethane-1,2-diyl)bis(N2-((E)-3-phenylallylidene)ethane-1,2-diamine)) was prepared by sonochemical process and characterized by single crystal X-ray crystallography, scanning electron microscopy (SEM), FT-IR and NMR spectroscopy and X-ray powder diffraction (XRPD). The X-ray

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service