Propionic acid
Synonym(s):
Acid C3, Propanoic acid, Propanyl acid
Linear Formula:
CH3CH2COOH
CAS Number:
Molecular Weight:
74.08
Products
Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Analyte Suitability
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing