Skip to Content
Merck
All Photos(2)

Key Documents

36168

Supelco

MGK 264

PESTANAL®, analytical standard

Synonym(s):

N-(2-Ethylhexyl)-5-norbornene-2,3-dicarboximide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H25NO2
CAS Number:
Molecular Weight:
275.39
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

technical mixture of isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCCCC(CC)CN1C(=O)C2[C@H]3C[C@H](C=C3)C2C1=O

InChI

1S/C17H25NO2/c1-3-5-6-11(4-2)10-18-16(19)14-12-7-8-13(9-12)15(14)17(18)20/h7-8,11-15H,3-6,9-10H2,1-2H3/t11?,12-,13+,14?,15?

InChI key

WLLGXSLBOPFWQV-OTHKPKEBSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

MGK 264 is both a synergist and a mosquito repellent.

Application

MGK 264 may be used as an analytical reference standard for the determination of the target analyte in pyrethrum extracts and indoor air by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Skin Sens. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

350.6 °F - closed cup

Flash Point(C)

177 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stir bar sorptive extraction coupled to thermodesorption-gas chromatography-mass spectrometry for the determination of insect repelling substances in water samples.
Rodil R and Moeder M
Journal of Chromatography A, 1178(1-2), 9-16 (2008)
Simultaneous determination of semivolatile organic compounds in indoor air by gas chromatography-mass spectrometry after solid-phase extraction.
Yoshida T, et al.
Journal of Chromatography A, 1023(2), 255-269 (2004)
H S Radwan et al.
Toxicology, 14(3), 193-198 (1979-11-01)
Piperonyl butoxide, sulfoxide and MGK 264 were found to act as synergists for certain organotin compounds; with respect to their toxicity on the larvae of a susceptible strain of the cotton leafworm, Spodoptera littoralis (Boisd.). High degree of synergism was
Priyamvada Singh et al.
Pest management science, 61(2), 204-208 (2004-12-28)
The effects of sub-lethal treatments (20 and 60% of 24-h LC(50)) with plant-derived molluscicides Annona squamosa, acetogenins, Argemone mexicana seed and protopine, in combination (1 + 5) with MGK-264 (ENT 8184) or piperonyl butoxide on the reproduction of Lymnaea acuminata
I H Wang et al.
Journal of chromatography. A, 864(2), 271-281 (2000-02-11)
N-Octyl bicycloheptene dicarboximide (MGK 264) has exo and endo diastereomers. Each structure has a chiral center at the nitrogen side chain. Enantioselective separation of MGK 264 was achieved by normal-phase high-performance liquid chromatography (HPLC) using cellulose-based Chiralcel OD column with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service