Skip to Content
Merck
All Photos(1)

Key Documents

34135

Supelco

Deepoxy-deoxynivalenol solution

~50 μg/mL in acetonitrile, analytical standard

Synonym(s):

3α,7α,15-Trihydroxy-trichothec-9,12-dien-8-one solution, DOM-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O5
CAS Number:
Molecular Weight:
280.32
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

~50 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

CC1=C[C@H]2O[C@@H]3[C@H](O)C[C@@](C)(C3=C)[C@@]2(CO)[C@H](O)C1=O

InChI

1S/C15H20O5/c1-7-4-10-15(6-16,13(19)11(7)18)14(3)5-9(17)12(20-10)8(14)2/h4,9-10,12-13,16-17,19H,2,5-6H2,1,3H3/t9-,10-,12+,13-,14+,15-/m1/s1

InChI key

ZACLXWTWERGCLX-MDUHGFIHSA-N

General description

Certan Vial
Deepoxy-deoxynivalenol is found to be a metabolite of the trichothecene mycotoxin deoxynivalenol, which can be generally found in swine.

Application

Deepoxy-deoxynivalenol solution may be used as internal standard in the determination of trichothecene mycotoxins present in baby foods, using high performance liquid chromatography coupled with mass spectrometry (HPLC/MS-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥92.5% (HPLC)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Zhu et al.
Journal of chromatography. A, 1556, 81-87 (2018-05-08)
Deoxynivalenol (DON) is a type B trichothecene mycotoxin that is commonly detected in grains infested with Fusarium species. The maximum tolerated levels of DON in the majority of world's countries are restricted to 0.75 mg kg-1 within the human food chain and
Sandra Debevere et al.
Toxins, 12(2) (2020-02-09)
Ruminants are generally considered to be less susceptible to the effects of mycotoxins than monogastric animals as the rumen microbiota are capable of detoxifying some of these toxins. Despite this potential degradation, mycotoxin-associated subclinical health problems are seen in dairy
Arnau Vidal et al.
Food research international (Ottawa, Ont.), 101, 139-147 (2017-09-25)
Deoxynivalenol (DON) is one of the most frequently occurring mycotoxins in wheat crops worldwide and poses a risk to human and animal health due to its wide range of adverse effects. Deoxynivalenol-3-glucoside (DON-3-glucoside) is a DON plant conjugate that is
Xiaojuan Gao et al.
Toxins, 12(2) (2020-01-30)
Deoxynivalenol (DON) is a highly abundant mycotoxin that exerts many adverse effects on humans and animals. Much effort has been made to control DON in the past, and bio-transformation has emerged as the most promising method. However, useful and effective
Nathan Broekaert et al.
Archives of toxicology, 91(2), 699-712 (2016-04-22)
Crossover animal trials were performed with intravenous and oral administration of deoxynivalenol-3-β-D-glucoside (DON3G) and deoxynivalenol (DON) to broiler chickens and pigs. Systemic plasma concentrations of DON, DON3G and de-epoxy-DON were quantified using liquid chromatography-tandem mass spectrometry. Liquid chromatography coupled to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service