Skip to Content
Merck
All Photos(1)

Key Documents

32911

Supelco

Deoxynivalenol 3-glucoside solution

~50 μg/mL in acetonitrile, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O11
CAS Number:
Molecular Weight:
458.46
Beilstein:
5462389
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

~50 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

CC1=C[C@H]2O[C@@H]3[C@@H](C[C@@](C)([C@]34CO4)[C@@]2(CO)[C@H](O)C1=O)O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O

InChI

1S/C21H30O11/c1-8-3-11-20(6-23,16(28)12(8)24)19(2)4-9(17(32-11)21(19)7-29-21)30-18-15(27)14(26)13(25)10(5-22)31-18/h3,9-11,13-18,22-23,25-28H,4-7H2,1-2H3/t9-,10-,11-,13-,14+,15-,16-,17-,18-,19-,20-,21+/m1/s1

InChI key

PUMXWMGECQIOGB-SMSDQXDJSA-N

General description

Certan Vial
Check out our entire range of trichothecene type mycotoxin reference materials

Deoxynivalenol 3-glucoside solution is the major metabolite of deoxynivalenol (DON) and a modified mycotoxin. The conjugation of deoxynivalenol (DON) with glucose formed by the metabolism of plants results in the formation of deoxynivalenol-3-glucoside.

Application

Deoxynivalenol 3-glucoside may be used as a reference standard for the determination of deoxynivalenol 3-glucoside in wheat and barley by liquid chromatography coupled to mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥92.5% (HPLC)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromatographic methods for the simultaneous determination of mycotoxins and their conjugates in cereals.
Berthiller F, et al.
International Journal of Food Microbiology, 119(1-2), 33-37 (2007)
Determination of deoxynivalenol and deoxynivalenol-3-glucoside in wheat and barley using liquid chromatography coupled to mass spectrometry: on-line clean-up versus conventional sample preparation techniques.
Nathanail AV, et al.
Journal of Chromatography A, 1374(1-2), 31-39 (2014)
Simultaneous determination of deoxynivalenol, deoxynivalenol-3-glucoside and nivalenol in wheat grains by HPLC-PDA with immunoaffinity column cleanup.
Trombete F, et al.
Food Analytical Methods, 9(9), 2579-2586 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service