Skip to Content
Merck
All Photos(1)

Key Documents

34133

Supelco

15-Acetyldeoxynivalenol solution

~100 μg/mL in acetonitrile, analytical standard

Synonym(s):

15-ADON, 15-AcDON, 15-Acetoxy-3α,7α-dihydroxy-12,13-epoxytrichothec-9-en-8-one, 15-Acetylvomitoxin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22O7
CAS Number:
Molecular Weight:
338.35
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

~100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

[H][C@]12O[C@]3([H])[C@H](O)C[C@@](C)([C@]34CO4)[C@@]1(COC(C)=O)[C@H](O)C(=O)C(C)=C2

InChI

1S/C17H22O7/c1-8-4-11-16(6-22-9(2)18,13(21)12(8)20)15(3)5-10(19)14(24-11)17(15)7-23-17/h4,10-11,13-14,19,21H,5-7H2,1-3H3/t10-,11-,13-,14-,15-,16-,17+/m1/s1

InChI key

IDGRYIRJIFKTAN-HTJQZXIKSA-N

General description

Certan Vial
Deoxynivalenol (DON) has a 12, 13 - epoxy group, three OH functions, and an a,ß-unsaturated keto group.[1] It is produced by various Fusarium spp.[2] It is slightly soluble in water but highly soluble in polar solvents such as methanol, ethyl acetate and acetonitrile and is stable in organic solvents specially ethyl acetate and acetonitrile therefore making them highly suitable for storage.[1] 15-Acetyldeoxynivalenol (15-AcDON) is a low molecular weight sesquiterpenoid trichothecene mycotoxin that basically means it is a monoacetyl analogue of DON.[3] Furthermore it is one of the most common mycotoxin present in food and animal feed. [4]

Application

15-Acetyldeoxynivalenol solution was used as a reference to develop enzyme immunoassay (EIA) techniques as one of the screening methods for 15-AcDon and its application in wheat.[5]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥97.0% (HPLC)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meeting the Mycotoxin Menace.
Barug D.
Science, 255-256 (2004)
Safety Evaluation of Certain Mycotoxins in Food.
Henry SH, et al.
Technology & Engineering, 471-472 (2001)
Production of monoclonal antibodies for the specific detection of deoxynivalenol and 15-acetyldeoxynivalenol by ELISA.
Sinha RC, et al.
Journal of Agricultural and Food Chemistry, 43 (6), 1740-1744 (1995)
Two formats of enzyme immunoassay for 15-acetyldeoxynivalenol applied to wheat.
Usleber E, et al.
Journal of Agricultural and Food Chemistry, 41 (11), 2019-2023 (1993)
María Marta Reynoso et al.
International journal of food microbiology, 145(2-3), 444-448 (2011-02-16)
Argentina is the fourth largest exporter of wheat in the world. The main pathogen associated with Fusarium Head Blight (FHB) of wheat in Argentina is Fusarium graminearum lineage 7 also termed F. graminearum sensu stricto in the F. graminearum species

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service