Skip to Content
Merck
All Photos(2)

Key Documents

34058

Supelco

Norfloxacin-d5

VETRANAL®, analytical standard

Synonym(s):

6-Fluoro-1,4-dihydro-4-oxo-1-pentadeuteroethyl-7-piperazino-3-quinolinecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13D5FN3O3
CAS Number:
Molecular Weight:
324.36
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental
forensics and toxicology
pharmaceutical (small molecule)

format

neat

mass shift

M+5

SMILES string

[2H]C([2H])([2H])C([2H])([2H])N1C=C(C(O)=O)C(=O)c2cc(F)c(cc12)N3CCNCC3

InChI

1S/C16H18FN3O3/c1-2-19-9-11(16(22)23)15(21)10-7-12(17)14(8-13(10)19)20-5-3-18-4-6-20/h7-9,18H,2-6H2,1H3,(H,22,23)/i1D3,2D2

InChI key

OGJPXUAPXNRGGI-ZBJDZAJPSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shengnan Zhao et al.
The Science of the total environment, 569-570, 1350-1358 (2016-07-09)
As special zones, the intertidal zones of the Yellow River Delta (YRD) are highly variable along with time and space. Fluvial-marine and land-ocean interactions which frequently occur in these areas have a great impact on the fate of pollutants. Antibiotics
Chao Song et al.
Chemosphere, 161, 127-135 (2016-07-18)
Antibiotics are widely used to improve the health and yields of farmed animals, including fish, but their use is accompanied by undesirable ecological effects. Relatively little is known about the water-body burden of antibiotics and their influence on primary productivity
Pinjing He et al.
Journal of environmental sciences (China), 85, 17-34 (2019-09-01)
Is our food safe and free of the crisis of antibiotics and antibiotic resistance (AR)? And will the derived food waste (FW) impose AR risk to the environment after biological treatment? This study used restaurant FW leachates flowing through a
Min Hu et al.
Journal of agricultural and food chemistry, 69(5), 1656-1666 (2021-01-28)
It is commonly known that the widespread use of antibiotics has led to their existence in food products as residues and ingestion of these food products may create a selection pressure on bacteria inhabiting the human body. In this study
Fangkai Zhao et al.
Chemosphere, 219, 882-895 (2018-12-21)
Long-term manure application gives rise to the uptake of antibiotics by plants and antibiotics subsequent entry into the food chain, representing an important alternative pathway for human exposure to antibiotics. The antibiotics can cause negative effects on crop growth and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service