Skip to Content
Merck
All Photos(1)

Key Documents

33654

Supelco

Spirodiclofen

PESTANAL®, analytical standard

Synonym(s):

3-(2,4-Dichlorophenyl)-2-oxo-1-oxaspiro[4.5]dec-3-en-4-yl) 2,2-dimethylbutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24Cl2O4
CAS Number:
Molecular Weight:
411.32
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCC(C)(C)C(=O)OC1=C(C(=O)OC12CCCCC2)c3ccc(Cl)cc3Cl

InChI

1S/C21H24Cl2O4/c1-4-20(2,3)19(25)26-17-16(14-9-8-13(22)12-15(14)23)18(24)27-21(17)10-6-5-7-11-21/h8-9,12H,4-7,10-11H2,1-3H3

InChI key

DTDSAWVUFPGDMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Spirodiclofen is a selective, non-systemic acaricide from the new chemical class of tetronic acid derivatives. It shows activity against all developmental stages of female adults of tetranychid mite species such as the two-spotted spider miteTetranychus urticae.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Spirodiclofen may be used as analytical reference standard for the determination of the pesticide, spirodiclofen in crops using high-performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS).

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Carc. 1B - Repr. 2 - Skin Sens. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic and biochemical analysis of a laboratory-selected spirodiclofen-resistant strain of Tetranychus urticae Koch (Acari: Tetranychidae).
Pottelberge VS, et al.
Pest Management Science, 65(4), 358-366 (2009)
Spirodiclofen resistance risk assessment in Tetranychus urticae (Acari: Tetranychidae): a biochemical approach.
Rauch N and Nauen R
Pesticide Biochemistry and Physiology, 74(2), 91-101 (2002)
Simultaneous analysis of 70 pesticides using HPLC/MS/MS: a comparison of the multiresidue method of Klein and Alder and the QuEChERS method.
Riedel M and Speer K
Journal of AOAC (Association of Official Analytical Chemists) International, 93(6), 1972-1986 (2010)
Nicky Wybouw et al.
Genetics, 211(4), 1409-1427 (2019-02-13)
Pesticide resistance arises rapidly in arthropod herbivores, as can host plant adaptation, and both are significant problems in agriculture. These traits have been challenging to study as both are often polygenic and many arthropods are genetically intractable. Here, we examined
Dandan Xu et al.
Pesticide biochemistry and physiology, 150, 89-96 (2018-09-10)
The two-spotted spider mite, Tetranychus urticae Koch, is a serious agricultural pest that has developed resistance to many pesticides. A leaf dip assay was used to assess the resistance of seven field populations of T. urticae to 11 pesticides in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service