Skip to Content
Merck
All Photos(1)

Key Documents

33152

Supelco

1,5-Diphenylcarbazide

reag. Ph. Eur., ≥98.0%

Synonym(s):

1,5-Diphenylcarbohydrazide, sym.-Diphenylcarbazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHNHCONHNHC6H5
CAS Number:
Molecular Weight:
242.28
Beilstein:
752039
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
reag. Ph. Eur.

Quality Level

Assay

≥98% (HPLC)
≥98.0%

technique(s)

UV/Vis spectroscopy: suitable
titration: suitable

impurities

diphenylcarbazone, in accordance
≤0.1% insoluble matter in ethanol

ign. residue

≤0.05% (as SO4)

mp

170-175 °C (lit.)
170-175 °C

suitability

passes test for Hg detection

SMILES string

O=C(NNc1ccccc1)NNc2ccccc2

InChI

1S/C13H14N4O/c18-13(16-14-11-7-3-1-4-8-11)17-15-12-9-5-2-6-10-12/h1-10,14-15H,(H2,16,17,18)

InChI key

KSPIHGBHKVISFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,5-Diphenylcarbazide reaction with chromium(VI), has been widely used for the spectro- photometric determination of chromium. It is also an electron donor, and can be used for measuring photosystem 2 activity in subchloroplast fragments.

Application

1,5-diphenylcarbazide was used for preparing diphenylcarbazide solution, during an experimental procedure done for reducing biological chromium(VI), using trickling filter. It may be used for pre-column complexation in HPLC, during on-line preconcentration and determination of chromium(VI) traces in water.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biological chromium (VI) reduction using a trickling filter.
Dermou E
Journal of Hazardous Materials, 126 (1), 78-85 (2005)
On-line preconcentration and determination of chromium (VI) in waters by high-performance liquid chromatography using pre-column complexation with 1, 5-diphenylcarbazide.
Padarauskas A
Journal of Chromatography A, 808 (1), 193-199 (1998)
Oxidation of 1, 5-diphenylcarbazide as a measure of photosystem 2 activity in subchloroplast fragments.
Vernon LP and Shaw ER.
Biochemical and Biophysical Research Communications, 36 (6), 878-884 (1969)
N Rajesh et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 69(2), 612-618 (2007-07-03)
A novel approach has been developed for the solid phase extraction of chromium(VI) based on the adsorption of its diphenylcarbazide complex on a mixture of acid activated montmorillonite (AAM)-silica gel column. The effect of various parameters such as acidity, stability
M L Ghirardi et al.
Biochemistry, 35(6), 1820-1828 (1996-02-13)
The inhibition of DPC-mediated DCIP photoreduction by exogenous MnCl2 in Tris-treated photosystem II (PSII) membrane fragments has been used to probe for amino acids on the PSII reaction center proteins, including D1His337, that provide ligands for binding manganese [Preston, C.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service