Skip to Content
Merck
All Photos(2)

Key Documents

D194255

Sigma-Aldrich

1,3-Dinitrobenzene

97% anhydrous basis

Synonym(s):

1,3-Dinitrobenzene, DNB, Meta-dinitrobenzene, m-Dinitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(NO2)2
CAS Number:
Molecular Weight:
168.11
Beilstein:
1105654
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97% anhydrous basis

form

solid

impurities

≤10.0% H2O

bp

297 °C (lit.)

mp

84-86 °C (lit.)

density

1.368 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1cccc(c1)[N+]([O-])=O

InChI

1S/C6H4N2O4/c9-7(10)5-2-1-3-6(4-5)8(11)12/h1-4H

InChI key

WDCYWAQPCXBPJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Dinitrobenzene can undergo reductive degradation by alkaline ascorbic acid to produce 1,3-phenylenediamine.

Application

1,3-Dinitrobenzene can be used as a reactant to synthesize:      
  • 2′,6′- dinitrobiphenyl-4-ol and 1-nitrodibenzofuran via copper-catalyzed regioselective cross-coupling reaction with 4-iodophenol and 2-iodophenol respectively.      
  • 1H-indazole derivatives by reacting with various N-tosylhydrazones in the presence of a base catalyst.      
  • 3-(1H-Tetrazol-1-yl)benzenamine by reacting with triethyl orthoformate and NaN3 in the presence of a three-functional redox catalytic system.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidation/MCR domino protocol for direct transformation of methyl benzene, alcohol, and nitro compounds to the corresponding tetrazole using a three-functional redox catalytic system bearing TEMPO/Co (III)-porphyrin/Ni (II) complex
Mahmoudi B, et al.
Molecular Catalysis, 499, 111311-111311 (2021)
Zhenxing Liu et al.
Chemical communications (Cambridge, England), 50(39), 5061-5063 (2014-04-10)
A new method for the synthesis of 1H-indazoles from readily available N-tosylhydrazones and nitroaromatic compounds has been developed. This transformation occurs under transition-metal-free conditions and shows a wide substrate scope. The method has been successfully applied to the formal synthesis
1, 3-Dinitrobenzene reductive degradation by alkaline ascorbic acid--Reaction mechanisms, degradation pathways and reagent optimization
Ciou C,et al.
Chemosphere, 166, 482-488 (2017)
Copper-Mediated Vicarious Substitution of 1, 3-Dinitrobenzene with lodophenols or lodomethyl Phenyl Sulfoxide/Sulfone
Haglund O, et al.
Synthesis, 1990(10), 942-944 (1990)
Chenyu Lin et al.
BMC chemistry, 14(1), 66-66 (2020-12-10)
For analysis of weak π-π complexes proton-nuclear magnetic resonance (proton-NMR) simultaneously provides information of stacking configurations and association constants [Formula: see text] However, an apparent issue for this approach is inconsistent/impossible constant estimation which often leads to unreasonable interpretation for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service