Skip to Content
Merck
All Photos(1)

Key Documents

32882

Supelco

4-tert-Octylphenol monoethoxylate solution

10 μg/mL in acetone, analytical standard

Synonym(s):

2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H26O2
CAS Number:
Molecular Weight:
250.38
Beilstein:
2114647
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

−20°C

SMILES string

CC(C)(C)CC(C)(C)c1ccc(OCCO)cc1

InChI

1S/C16H26O2/c1-15(2,3)12-16(4,5)13-6-8-14(9-7-13)18-11-10-17/h6-9,17H,10-12H2,1-5H3

InChI key

JYCQQPHGFMYQCF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sigma-Aldrich offers 4-tert-octylphenol monoethoxylate solution for a variety of applications that include, but are not limited to, environmental, food and beverage, forensics, petrochemical and pharmaceutical analyzes.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie Orre et al.
Neurobiology of aging, 35(12), 2746-2760 (2014-07-09)
Reactive astrocytes and microglia are associated with amyloid plaques in Alzheimer's disease (AD). Yet, not much is known about the molecular alterations underlying this reactive phenotype. To get an insight into the molecular changes underlying AD induced astrocyte and microglia
Pasqualino De Antonellis et al.
Molecular & cellular proteomics : MCP, 13(8), 2114-2131 (2014-06-11)
Several genes encoding for proteins involved in proliferation, invasion, and apoptosis are known to be direct miR-34a targets. Here, we used proteomics to screen for targets of miR-34a in neuroblastoma (NBL), a childhood cancer that originates from precursor cells of
Nobuhiko Hamazaki et al.
Development (Cambridge, England), 142(5), 910-920 (2015-01-31)
In mice, zygotic activation occurs for a wide variety of genes, mainly at the 2-cell stage. Long noncoding RNAs (lncRNAs) are increasingly being recognized as modulators of gene expression. In this study, directional RNA-seq of MII oocytes and 2-cell embryos
Olga Pougovkina et al.
Human molecular genetics, 23(13), 3513-3522 (2014-02-12)
Mitochondria integrate metabolic networks for maintaining bioenergetic requirements. Deregulation of mitochondrial metabolic networks can lead to mitochondrial dysfunction, which is a common hallmark of many diseases. Reversible post-translational protein acetylation modifications are emerging as critical regulators of mitochondrial function and
Matasha Dhar et al.
Molecular endocrinology (Baltimore, Md.), 28(7), 1073-1087 (2014-06-01)
Leptin acts in the hippocampus to enhance cognition and reduce depression and anxiety. Cognitive and emotional disorders are associated with abnormal hippocampal dendritic spine formation and synaptogenesis. Although leptin has been shown to induce synaptogenesis in the hypothalamus, its effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service