Skip to Content
Merck
All Photos(1)

Key Documents

31550

Supelco

2,3-Butanedione monoxime

for spectrophotometric det. of urea, ≥99.0%

Synonym(s):

BDM, Biacetyl monoxime, Diacetyl monoxime

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(=NOH)COCH3
CAS Number:
Molecular Weight:
101.10
Beilstein:
605582
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥98.0% (N)
≥99.0%

form

solid

quality

for spectrophotometric det. of urea

technique(s)

UV/Vis spectroscopy: suitable

ign. residue

≤0.05% (as SO4)

bp

185-186 °C (lit.)

mp

75-76 °C
75-78 °C (lit.)

SMILES string

CC(=O)\C(C)=N\O

InChI

1S/C4H7NO2/c1-3(5-7)4(2)6/h7H,1-2H3/b5-3+

InChI key

FSEUPUDHEBLWJY-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3-Butanedione Monoxime (BDM) is also known as diacetyl monoxime, it is also a nucleophilic agent and it can dephosphorylates acetylcholinesterase poisoned with organophosphates. It is also a well-characterized, non-competitive inhibitor of skeletal muscle myosin-II, where it inhibits the chemical and motile activity.

Application


  • A novel method to extend viability and functionality of living heart slices.: This research introduces a novel application of 2,3-Butanedione monoxime for prolonging the functional lifespan of cardiac tissue samples in experimental settings, offering insights into cardiac biology and potential therapeutic targets (Ross et al., 2023).

  • Molecular Mechanisms of Deregulation of Muscle Contractility Caused by the R168H Mutation in TPM3 and Its Attenuation by Therapeutic Agents.: The study utilizes 2,3-Butanedione monoxime to investigate the molecular pathways affected by genetic mutations in muscle contractility, contributing to the understanding of muscle disorders and their management (Karpicheva et al., 2023).

  • Generation of myocyte agonal Ca(2+) waves and contraction bands in perfused rat hearts following irreversible membrane permeabilisation.: Research employing 2,3-Butanedione monoxime investigates its role in inducing specific cellular events in cardiac cells under stress, highlighting its potential in studies of heart disease mechanisms and therapies (Morishita et al., 2023).


Analysis Note

Solubility:
0.5 g are completely soluble and give a clear solution in 10 mL water or also in 10 mL ethanol.
Sensitivity test:
0.05 mg urea in 3 mL water and 5 mL conc. HCl together with a 3% solution in water of diacetylmonoxime heated on a water bath for 10 minutes give a light yellow color.

Other Notes

Reagent for the colorimetric determination of urea and ureido-compounds; Spectrometric reagent for Co(II), Ni(II), Pd(II) and Re(VII); Reagent for the gravimetric determination of Ni(II)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2, 3-Butanedione monoxime (BDM) as a myosin inhibitor.
Ostap EM.
Journal of Muscle Research and Cell Motility, 23 (4), 305-308 (2002)
Multiple Effects of 2, 3-Butanedione Monoxime.
Sellin LC and McArdle JJ/
Pharmacology & Toxicology, 74 (4-5), 305-313 (1994)
Improvements on the Prescott-Jones method for the colorimetric analysis of ureido compounds.
D B Shindler et al.
Analytical biochemistry, 97(2), 421-422 (1979-09-01)
W.B. Guenther
Analytical Letters, 12A, 1305-1305 (1979)
Chang Liu et al.
Molecular plant, 11(11), 1389-1399 (2018-10-09)
The process of pollen germination is crucial for flowering plant reproduction, but the mechanisms through which pollen grains establish polarity and select germination sites are not well understood. In this study, we report that a formin family protein, AtFH5, is localized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service