Skip to Content
Merck
All Photos(1)

Key Documents

130060

Sigma-Aldrich

4-Methyl-3-thiosemicarbazide

97%

Synonym(s):

N-Methylhydrazinecarbothioamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NHCSNHNH2
CAS Number:
Molecular Weight:
105.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

135-138 °C (lit.)

functional group

amine

SMILES string

CNC(=S)NN

InChI

1S/C2H7N3S/c1-4-2(6)5-3/h3H2,1H3,(H2,4,5,6)

InChI key

PTVZQOAHCSKAAS-UHFFFAOYSA-N

General description

4-Methyl-3-thiosemicarbazide is a subject of considerable pharmacological interest owing to its wide range of biological effects.

Application

4-Methyl-3-thiosemicarbazide was used in the analysis of electronic features of sulfur-using, high-resolution, low-temperature X-ray diffraction datasets.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electronic features and hydrogen bonding capacity of the sulfur acceptor in thioureido-based compounds. Experimental charge density study of 4-methyl-3-thiosemicarbazide.
Francuski BM, et al. et al.
CrystEngComm, 13, 3580-3591 (2011)
D Zahn et al.
Water research, 150, 86-96 (2018-12-06)
Transformation of industrial chemicals might be a significant source of hitherto unknown persistent and mobile organic contaminants (PMOC, PM chemicals) present in the aquatic environment. Herein we depicted a three-step strategy consisting of (I) the prioritization of potential PMOC precursors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service