Skip to Content
Merck
All Photos(2)

Key Documents

298409

Sigma-Aldrich

Uniblue A sodium salt

Reactive stain for proteins, ≥70% (HPLC)

Synonym(s):

Acid Blue 215

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H15N2NaO7S2
CAS Number:
Molecular Weight:
506.48
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

≥70% (HPLC)

form

powder

mp

>300 °C (lit.)

λmax

596 nm

εmax

≥20000 at 253-259 nm in water at 0.01 g/L
≥23000 at 224-230 nm in water at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Nc1c2C(=O)c3ccccc3C(=O)c2c(Nc4cccc(c4)S(=O)(=O)C=C)cc1S([O-])(=O)=O

InChI

1S/C22H16N2O7S2.Na/c1-2-32(27,28)13-7-5-6-12(10-13)24-16-11-17(33(29,30)31)20(23)19-18(16)21(25)14-8-3-4-9-15(14)22(19)26;/h2-11,24H,1,23H2,(H,29,30,31);/q;+1/p-1

InChI key

ANOULJGZTVOIFB-UHFFFAOYSA-M

Application

Reactive stain for proteins

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Amorisco et al.
Rapid communications in mass spectrometry : RCM, 25(13), 1801-1811 (2011-06-04)
The structural elucidation of carbonyl-containing by-products arising from Uniblu-OH ozonation has been investigated by liquid chromatography/electrospray ionization tandem mass spectrometry (LC/ESI-MS/MS) employing a quadrupole time-of-flight mass spectrometer. The by-products were derivatized with 2,4-dinitrophenylhydrazine, allowing the formation of [M-H](-) ions of
Marco A Mata-Gómez et al.
PloS one, 7(2), e31438-e31438 (2012-03-01)
The identification of proteins by mass spectrometry is a standard method in biopharmaceutical quality control and biochemical research. Prior to identification by mass spectrometry, proteins are usually pre-separated by electrophoresis. However, current protein staining and de-staining protocols are tedious and
R Bültmann et al.
Naunyn-Schmiedeberg's archives of pharmacology, 359(4), 339-344 (1999-05-27)
In order to assess the consequences of a concomitant blockade of P2X-receptors and ecto-nucleotidases, effects of 13 P2-receptor antagonists were investigated on contractions of the rat vas deferens elicited by alpha,beta-methylene ATP (alpha,beta-MeATP) and ATP and on the removal of
F Herrera et al.
Water research, 35(3), 750-760 (2001-03-07)
Experimental results from the adsorption and subsequent catalytic combustion of the reactive dye Uniblue A on hematite indicate that this iron oxide can be used as an affordable catalyst for environmental purposes. Uniblue A was adsorbed on hematite and the
G Paramaguru et al.
Journal of hazardous materials, 175(1-3), 985-991 (2009-11-27)
The interaction between lysozyme and anthraquinone dyes such as Alizarin Red S, Acid blue 129 and Uniblue was studied using steady state, time resolved fluorescence measurements and docking studies. Addition of anthraquinone dyes effectively quenched the intrinsic fluorescence of lysozyme.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service