28632-U
Hamilton® 1701N syringe
1701N Slim Line, volume 10 μL, needle size 22s ga (blunt tip), needle L 51 mm (2 in.), pkg of 1 ea
About This Item
Recommended Products
material
blunt tip stainless steel (point style 3)
needle size
22s ga (blunt tip)
description
gastight, 10uL
feature
GASTIGHT® Syringe, Fixed needle
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 203194
needle L
51 mm (2 in.)
volume
10 μL
compatibility
for use with CTC HTS PAL Instruments (HPLC sampling)
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service