Key Documents
26208
Hamilton® syringe, 1000 series GASTIGHT®, PTFE luer lock
1001TLL, PTFE Luer lock (with slots), volume 1 mL, needle size (not included)
About This Item
UNSPSC Code:
41122001
Pricing and availability is not currently available.
material
PTFE Luer lock (with slots)
needle size
(not included)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81327
needle L
, n/a
volume
1 mL
fitting
luer lock (without slots)
compatibility
for use with Hamilton 1001TLL (PTFE Luer lock)
General description
Hamilton syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed.[1] The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.[2]
Application
It has been used in an experiment performed to inject magnetic beads into Lab-on-a-Chip system in order to demonstrate controlled movement of the magnetic beads within a microfluidic channel.[3] It was also used in alkene termination through olefin cross-metathesis reaction performed with gallium nitride wafers.[4]
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 06-Dec, 192-192 (2012)
Controlled counter-flow motion of magnetic bead chains rolling along microchannels.
Karle, Marc, et al.
Microfluidics and nanofluidics, 10.4, 935-939 (2011)
Pharmacology for the Prehospital Professional, 54-54 (2009)
Covalent attachment of a peptide to the surface of gallium nitride.
Makowski, Matthew S., et al.
Surface Science, 605.15, 1466-1475 (2011)
Jeffrey S. Guy
Pharmacology for the Prehospital Professional, 54-54 (2009)