Skip to Content
Merck
All Photos(2)

Key Documents

23701

Supelco

Acrylamide

analytical standard

Synonym(s):

2-Propenamide, Acrylic acid amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCONH2
CAS Number:
Molecular Weight:
71.08
Beilstein:
605349
EC Number:
MDL number:
UNSPSC Code:
12352111
eCl@ss:
39031205
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

2.45 (vs air)

vapor pressure

0.03 mmHg ( 40 °C)

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

125 °C/25 mmHg (lit.)

mp

82-86 °C (lit.)

solubility

water: soluble 200 g/L at 20 °C

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

NC(=O)C=C

InChI

1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)

InChI key

HRPVXLWXLXDGHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Acrylamide may be used as an analytical reference standard for the determination of the analyte in:
  • Environmental water samples using ion-exclusion chromatography-mass spectrometry system (IEC-MS).
  • Cereal-based food products by gas chromatography coupled to mass spectrometry (GC–MS) and reversed phase-high performance liquid chromatography with diode array detection (RP-HPLC-DAD).
  • Potato-based foods by liquid chromatography with diode array detection (LC–DAD).

Refer to the products certificate of analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trace level determination of acrylamide in cereal-based foods by gas chromatography- mass spectrometry.
Pittet A, et al.
Journal of Chromatography A, 1035(1), 123-130 (2004)
Cheremisinoff, P.
Handbook of Engineering Polymeric Materials, Tech & Eng, 61-61 (1997)
Determination of acrylamide in drinking water by large-volume direct injection and ion-exclusion chromatography- mass spectrometry.
Cavalli S, et al.
Journal of Chromatography A, 1039(1-2), 155-159 (2004)
RP-HPLC-DAD studies on acrylamide in cereal-based baby foods
Michalak J, et al.
J. Food Compos. Anal., 32, 68-73 (2013)
Determination of acrylamide in potato chips and crisps by high-performance liquid chromatography.
Gokmen V, et al.
Journal of Chromatography A, 1088(1-2), 193-199 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service