Skip to Content
Merck
All Photos(1)

Key Documents

01700

Sigma-Aldrich

Acrylamide

purum, ≥98.0% (GC)

Synonym(s):

2-Propenamide, Polyacrylamide, acrylic amide, akrylamid, Acrylic acid amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCONH2
CAS Number:
Molecular Weight:
71.08
Beilstein:
605349
EC Number:
MDL number:
UNSPSC Code:
41105319
eCl@ss:
39031205
PubChem Substance ID:
NACRES:
NB.22

vapor density

2.45 (vs air)

Quality Level

vapor pressure

0.03 mmHg ( 40 °C)

grade

purum

Assay

≥98.0% (GC)

bp

125 °C/25 mmHg (lit.)

mp

81-87 °C
82-86 °C (lit.)

solubility

water: soluble 200 g/L at 20 °C

storage temp.

room temp

SMILES string

NC(=O)C=C

InChI

1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)

InChI key

HRPVXLWXLXDGHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acrylamide has been used for preparing slab gel gradients for polyacrylamide gel electrophoresis (PAGE).

Biochem/physiol Actions

Acrylamide is a reactive, water soluble vinyl monomer. It causes a decrease in the number of neuritis per cell as well as reduces the rate of protein synthesis.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M W Cunningham et al.
The Journal of experimental medicine, 164(4), 998-1012 (1986-10-01)
mAbs produced by immunization of BALB/c mice with Streptococcus pyogenes M type 5 membranes were further characterized for their reaction with S. pyogenes pep M5 protein and with autoantigens associated with human cell lines. mAbs 36.2.2 and 54.2.8 simultaneously reacted
M Nordin-Andersson et al.
Cell biology and toxicology, 19(1), 43-51 (2003-03-29)
Basal cytotoxicity, morphological changes and alterations in cell physiological and neurochemical functions were studied in differentiated human neuroblastoma (SH-SY5Y) cells during exposure to acrylamide and during a subsequent recovery period after cessation of exposure. Acrylamide induced a 20% reduction in
Janneke G F Hogervorst et al.
Carcinogenesis, 35(5), 1032-1038 (2014-01-09)
Acrylamide, a probable human carcinogen, is present in heat-treated carbohydrate-rich foods. Epidemiological studies have not shown a clear association between acrylamide intake and colorectal cancer (CRC) risk. This may be due to the molecular heterogeneity in colorectal tumors, which was
Rusty L Montgomery et al.
EMBO molecular medicine, 6(10), 1347-1356 (2014-09-23)
Over the last decade, great enthusiasm has evolved for microRNA (miRNA) therapeutics. Part of the excitement stems from the fact that a miRNA often regulates numerous related mRNAs. As such, modulation of a single miRNA allows for parallel regulation of
Arianna Rath et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(39), 15668-15673 (2013-09-11)
SDS/PAGE is universally used in biochemistry, cell biology, and immunology to resolve minute protein amounts readily from tissue and cell extracts. Although molecular weights of water-soluble proteins are reliably determined from their SDS/PAGE mobility, most helical membrane proteins, which comprise

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service