Skip to Content
Merck
All Photos(1)

Key Documents

175633

Sigma-Aldrich

Silicone oil

high temperature

Synonym(s):

Poly(dimethylsiloxane-co-methylphenylsiloxane)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Si(CH3)2O]x[Si(CH3)(C6H5)O]y
CAS Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

<5 mmHg ( 25 °C)

Quality Level

parameter

−40-450 °F temp. range (−40-230 °C)

refractive index

n20/D 1.495

density

1.05 g/mL at 25 °C

SMILES string

C[Si](C)=O.C[Si](=O)c1ccccc1

InChI

1S/C7H8OSi.C2H6OSi/c1-9(8)7-5-3-2-4-6-7;1-4(2)3/h2-6H,1H3;1-2H3

InChI key

XWFOTLCDICUQTI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silicone oil is a highly viscous oil made up of poly(dimethylsiloxane-co-methylphenylsiloxane) chains.

Application

Silicone oil has been used as a medium component in containerless protein crystallization techniques which do not require contact with a solid surface. It was used as an oil bath during the synthesis of methyl 2-methylazulene-1-carboxylate.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

437.0 °F - closed cup

Flash Point(C)

225.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Colloidal ordering from phase separation in a liquid-crystalline continuous phase.
Loudet J-C, et al.
Nature, 407(6804), 611-613 (2000)
Hui-Ling Cao et al.
Acta crystallographica. Section D, Biological crystallography, 69(Pt 10), 1901-1910 (2013-10-09)
High-quality crystals are key to obtaining accurate three-dimensional structures of proteins using X-ray diffraction techniques. However, obtaining such protein crystals is often a challenge. Several containerless crystallization techniques have been reported to have the ability to improve crystal quality, but
Viscous liquid film flows over a periodic surface.
Trifonov YY.
International Journal of Multiphase Flow, 24(7), 1139-1161 (1999)
Jennifer Li et al.
The European journal of neuroscience, 34(12), 1983-1996 (2011-12-14)
Neuronal activity elicits metabolic and vascular responses, during which oxygen is first consumed and then supplied to the tissue via an increase in cerebral blood flow. Understanding the spatial and temporal dynamics of blood and tissue oxygen (To₂) responses following
Simon J Carter et al.
Xenobiotica; the fate of foreign compounds in biological systems, 50(4), 415-426 (2019-08-08)
Determine the inhibition mechanism through which cyclosporine inhibits the uptake and metabolism of atorvastatin in fresh rat hepatocytes using mechanistic models applied to data generated using a high throughput oil spin method.Atorvastatin was incubated in fresh rat hepatocytes (0.05-150 nmol/ml) with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service