Skip to Content
Merck
All Photos(1)

Key Documents

50384

Sigma-Aldrich

Silicone oil AP 150

viscosity ~150 mPa.s,   neat(25 °C)

Synonym(s):

Silicone fluid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

refractive index

n20/D 1.51

Quality Level

viscosity

~150 mPa.s, neat(25 °C)

General description

Silicone oil AP 150 is a conventional electrorheologic (ER) fluid. It was employed as an insulating medium during the poling of hard ferroelectric ceramics. Silicon oil is composed of a mixture of hydrophobic polymeric and monomeric compounds, containing silicon-oxygen bonds. Its physical and chemical characteristics have been reported. The utility of silicon oil as a vitreous substitute has been studied.

Application

Silicone oil AP 150 was used to prevent the water evaporation during rheological characterization of injectable polysaccharide system based on calcium alginate hydrogel and two dextran methacrylate derivatives. It may be used for the determination of chloroplast volume by the silicone oil filtering centrifugation technique.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

437.0 °F - closed cup

Flash Point(C)

225.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Aliotta et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 87(6), 062304-062304 (2013-07-16)
The influence of interactions between particle surface and host fluids in electrorheological suspensions is explored. It is observed that dispersions of nanosized particles of titania in octanoid acid exhibit an anomalously large electrorheologic effect when compared with a similar dispersion
Silicone Oil: Different Physical Proprieties and Clinical Applications.
Barca F, et al.
BioMed Research International, 2014 (2014)
S P Robinson
Plant physiology, 79(4), 996-1002 (1985-12-01)
Spinach leaf chloroplasts isolated in isotonic media (330 millimolar sorbitol, -1.0 megapascals osmotic potential) had optimum rates of photosynthesis when assayed at -1.0 megapascals. When chloroplasts were isolated in hypertonic media (720 millimolar sorbitol, -2.0 megapascals osmotic potential) the optimum
L Pescosolido et al.
European biophysics journal : EBJ, 39(6), 903-909 (2009-03-28)
A novel injectable polysaccharide system based on calcium Alginate (Ca-Alg) hydrogel and two Dextran methacrylate derivatives (DexMA) was recently developed. The resulting Interpenetrating Polymer Network showed a synergistic mechanical behavior that can be exploited to target the hydrogel properties towards
Poling of hard ferroelectric PZT ceramics.
Kamel TMand de With G.
J. Eur. Ceram. Soc., 28(9), 1827-1838 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service