Skip to Content
Merck
All Photos(1)

Key Documents

13468

Sigma-Aldrich

Sodium sulfide hydrate

≥60%, scales

Synonym(s):

μ-Sulfidedisodium hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na2S · xH2O
CAS Number:
Molecular Weight:
78.04 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

scales

Quality Level

reaction suitability

core: sodium
reagent type: catalyst

concentration

≥60%

anion traces

chloride (Cl-): ≤100 mg/kg

storage temp.

2-8°C

SMILES string

[Na]S[Na]

InChI

1S/2Na.S

InChI key

CXPWOVUZRAFMDA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

May contain up to 40% water.

Application

Nucleophile and reducing agent derived from H2S; reagent for the synthesis of thiols, thioethers, and acyclic sulfides, di-, and polysulfides; reduces aromatic nitro compounds to amines; reduces sulfoxides to sulfides.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total synthesis of xerulinic acid.
Achim Sorg et al.
Angewandte Chemie (International ed. in English), 43(34), 4523-4526 (2004-09-02)
Feixiang Zan et al.
Water research, 182, 115960-115960 (2020-07-06)
Volatile fatty acids (VFAs), the intermediate of the anaerobic process, are considered to be the critical, high-sensitive and reliable indicators of the process stability. Close monitoring and control of VFAs are paramount for the efficient operation of the anaerobic reactors.
Vanda Cerè et al.
The Journal of organic chemistry, 70(2), 664-669 (2005-01-18)
Optically pure enantiomers of the chiral tetrahydroxythiepane derivative 3,6-dihydroxy-4,5-O-isopropylidene-thiepane (3) are obtained using a novel protocol in which a library of all possible stereoisomers of 3 is synthesized, followed by two-step stereoselective chromatography, using, first, conventional achiral and, then, chiral
Krzysztof Walczyński et al.
European journal of medicinal chemistry, 40(1), 15-23 (2005-01-12)
In search for a new lead of non-imidazole histamine H3-receptor antagonists, a series of 1[(2-thiazolopyridine)-4-n-propyl]piperazines, the analogous 1-[(2-oxazolopyridine)-4-npropyl]piperazines, 1-[(2-benzothiazole)-4-n-propyl]piperazine and 1-[(2-benzooxazole)4-n-propyl]piperazine were prepared and in vitro tested as H3-receptor antagonists (the electrically evoked contraction of the guinea-pig jejunum). It appeared
Reza Sahraei et al.
Nanotechnology, 28(47), 475604-475604 (2017-10-12)
We report here on a one-pot, mild and low cost aqueous-based synthetic route for the preparation of colloidally stable and highly luminescent dual-doped Ag,Ni:ZnCdS/ZnS core/shell quantum dots (QDs). The pure dopant emission of the Ni-doped core/shell QDs was found to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service