Skip to Content
Merck
All Photos(1)

Key Documents

11365

Supelco

Avicel® PH-101 Cellulose Resin

285 mesh

Synonym(s):

Cellulose, Cellulose microcrystalline, Cellulose powder, Cotton linters

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

Product Name

Avicel® PH-101, ~50 μm particle size

form

powder

Quality Level

technique(s)

LPLC: suitable

matrix

Cellulose

matrix active group

polymer

particle size

~50 μm

separation technique

size exclusion (SEC)

SMILES string

O1[C@H](C(C(C(C1CO)O)O)O)O[C@@H]2C(OC(C(C2O)O)O)CO

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3?,4?,5?,6?,7?,8?,9?,10-,11?,12+/m1/s1

InChI key

GUBGYTABKSRVRQ-WFVLMXAXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Avicel® PH-101 was used as reference material in characterisation of microcrystalline cellulose (MCC) using Powder X-ray diffraction and Fourier transform infrared spectroscopy.
  • Avicel® PH-101 was used to evaluate crystallinity index and spectral analysis of cellulose using XRD and solid-state 13C NMR.
  • Avicel® PH-101 was used in preparation of phosphoric acid-swollen cellulose for enzyme assay using HPLC.
High purity cellulose powders for partition chromatography.

Legal Information

Avicel is a registered trademark of FMC Corp.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A staining technique for evaluating the pore structure variations of microcrystalline cellulose powders.
Yu, Xiaochun, and Rajai H. Atalla
Powder Technology, 98, 135-138 (1998)
Yasuya Fujita et al.
Applied and environmental microbiology, 70(2), 1207-1212 (2004-02-10)
A whole-cell biocatalyst with the ability to induce synergistic and sequential cellulose-degradation reaction was constructed through codisplay of three types of cellulolytic enzyme on the cell surface of the yeast Saccharomyces cerevisiae. When a cell surface display system based on
Physicochemical, spectroscopic and thermal properties of microcrystalline cellulose derived from corn cobs.
Azubuike, Chukwuemeka P., and Augustine O. Okhamafe
International Journal of Recycling of Organic Waste in Agriculture, 1, 1-7 (2012)
The physicochemical characteristic of microcrystalline cellulose, derived from sawdust, agricultural waste products
Oyeniyi, Y. J., and O. A. Itiola.
International Journal of Pharmacy and Pharmaceutical Sciences, 4, 197-200 (2012)
Sunkyu Park et al.
Biotechnology for biofuels, 3, 10-10 (2010-05-26)
Although measurements of crystallinity index (CI) have a long history, it has been found that CI varies significantly depending on the choice of measurement method. In this study, four different techniques incorporating X-ray diffraction and solid-state 13C nuclear magnetic resonance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service