Skip to Content
Merck
All Photos(2)

Key Documents

109584

Sigma-Aldrich

Pentyl acetate

99%

Synonym(s):

Amyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COO(CH2)4CH3
CAS Number:
Molecular Weight:
130.18
Beilstein:
1744753
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

4.5 (vs air)

Quality Level

vapor pressure

4 mmHg ( 20 °C)

Assay

99%

autoignition temp.

680 °F

refractive index

n20/D 1.402 (lit.)

bp

142-149 °C (lit.)

mp

−100 °C (lit.)

density

0.876 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCCCCOC(C)=O

InChI

1S/C7H14O2/c1-3-4-5-6-9-7(2)8/h3-6H2,1-2H3

InChI key

PGMYKACGEOXYJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentyl acetate is a straight chain ester. It is an odorant and its odor threshold has been evaluated by sensory panel methods. High pressure phase behavior of type I has been reported for its binary mixtures with CO2. Solubility of these solvent mixtures was observed to increase with the rise in temperature at constant pressure.

Application

Pentyl acetate may be used as odorant to compose the library of odorants to evaluate the responses of olfactory sensory neurons (OSNs) to various odorants (having different structures and odors).
Banana essence. Used as test odorant in studies of olfactory function and in studies of the psychosocial effects of odor.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

105.8 °F

Flash Point(C)

41 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kiyomitsu Nara et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 31(25), 9179-9191 (2011-06-24)
Mammals can perceive and discriminate myriad volatile chemicals as having a distinct odor. Odorants are initially detected by odorant receptors (ORs) on olfactory sensory neurons (OSNs) in the nose. In the mouse, each OSN expresses one of ∼1000 different OR
Odour thresholds of various branched and straight chain acetates.
Takeoka G, et al.
LWT--Food Science and Technology, 29(7), 667-680 (1996)
Janice K Kiecolt-Glaser et al.
Psychoneuroendocrinology, 33(3), 328-339 (2008-01-08)
Despite aromatherapy's popularity, efficacy data are scant, and potential mechanisms are controversial. This randomized controlled trial examined the psychological, autonomic, endocrine, and immune consequences of one purported relaxant odor (lavender), one stimulant odor (lemon), and a no-odor control (water), before
Jonas K Olofsson et al.
Chemical senses, 31(7), 699-704 (2006-07-20)
In experimental practice, odors are commonly applied to only one nostril for recordings of olfactory event-related potentials (OERPs), but the lateralization aspect of the OERP response is unclear regarding both stimulated nostril and cortical topography. The purpose of the present
Michael Schleyer et al.
Learning & memory (Cold Spring Harbor, N.Y.), 18(10), 639-653 (2011-09-29)
Drosophila larvae combine a numerically simple brain, a correspondingly moderate behavioral complexity, and the availability of a rich toolbox for transgenic manipulation. This makes them attractive as a study case when trying to achieve a circuit-level understanding of behavior organization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service