Merck
  • A behavior-based circuit model of how outcome expectations organize learned behavior in larval Drosophila.
A behavior-based circuit model of how outcome expectations organize learned behavior in larval Drosophila.
Learning & memory (Cold Spring Harbor, N.Y.) (2011-09-29)
Michael Schleyer, Timo Saumweber, Wiebke Nahrendorf, Benjamin Fischer, Désirée von Alpen, Dennis Pauls, Andreas Thum, Bertram Gerber
ABSTRACT
Drosophila larvae combine a numerically simple brain, a correspondingly moderate behavioral complexity, and the availability of a rich toolbox for transgenic manipulation. This makes them attractive as a study case when trying to achieve a circuit-level understanding of behavior organization. From a series of behavioral experiments, we suggest a circuitry of chemosensory processing, odor-tastant memory trace formation, and the "decision" process to behaviorally express these memory traces--or not. The model incorporates statements about the neuronal organization of innate vs. conditioned chemosensory behavior, and the types of interaction between olfactory and gustatory pathways during the establishment as well as the behavioral expression of odor-tastant memory traces. It in particular suggests that innate olfactory behavior is responsive in nature, whereas conditioned olfactory behavior is captured better when seen as an action in pursuit of its outcome. It incorporates the available neuroanatomical and behavioral data and thus should be useful as scaffold for the ongoing investigations of the chemo-behavioral system in larval Drosophila.
Sigma-Aldrich
Amyl acetate, ≥99%, FG
Sigma-Aldrich
Pentyl acetate, 99%
Sigma-Aldrich
Pentyl acetate, puriss. p.a., ≥98.5% (GC)