Skip to Content
Merck
All Photos(1)

Key Documents

10368

Supelco

trans-Anethole

analytical standard

Synonym(s):

4-Propenylanisole, trans-1-Methoxy-4-(1-propenyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHC6H4OCH3
CAS Number:
Molecular Weight:
148.20
Beilstein:
774229
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

contains

~0.05% 4-tert-butylcatechol as stabilizer

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.561 (lit.)
n20/D 1.561

bp

234-237 °C (lit.)

mp

20-21 °C (lit.)

density

0.988 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(\C=C\C)cc1

InChI

1S/C10H12O/c1-3-4-9-5-7-10(11-2)8-6-9/h3-8H,1-2H3/b4-3+

InChI key

RUVINXPYWBROJD-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-Anethole is a naturally occuring flavouring agent. It has insecticidal, larvicidal, and antimicrobial properties.

Application

It was used as standard in the determination of trans-anethole in Savia sclarea essential oil using HPLC and GC-MS method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Naturally occurring phenylpropene derivative that is estrogenic at lower concentrations and cytotoxic at higher concentrations to cancer cell lines. The cytotoxicity is related to the metabolism of trans-anethole to 4-hydroxy-1-propenylbenzene.

Other Notes

This compound is commonly found in plants of the genus: pimpinella

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

213.8 °F

Flash Point(C)

101 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of trans-anethole in Salvia sclarea essential oil by liquid chromatography and GC-MS.
Valgimiglib, Luca, Simone Gabbaninic, and Vanni Cavrinia.
Journal of Separation Science, 25, 703-709 (2002)
S K Abraham
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 39(5), 493-498 (2001-04-21)
The naturally occurring flavouring agents trans-anethole and eugenol were evaluated for antigenotoxic effects in mice. The test doses of trans-anethole (40-400 mg/kg body weight) and eugenol (50-500 mg/kg weight) were administered by gavage 2 and 20 h before the genotoxins
Bülent Cetin et al.
Journal of medicinal food, 13(1), 196-204 (2010-02-09)
The objective of this study was to determine the chemical compositions of the essential oil and hexane extract isolated from the inflorescence, leaf stems, and aerial parts of Florence fennel and the antimicrobial activities of the essential oil, hexane extract
Rajesh K Joshi et al.
Natural product communications, 6(1), 141-143 (2011-03-04)
The essential oil of the leaves of Feronia elephantum Corr. was analyzed by gas chromatography and gas chromatography/mass spectrometry. The main constituents were beta-pinene (28.4%), Z-anethole (22.1%), methyl chavicol (12.0%) and E-anethole (8.1%), among thirty-three identified compounds, which represented 92.6%
Eun Jeong Choo et al.
Biological & pharmaceutical bulletin, 34(1), 41-46 (2011-01-08)
Anethole is known to possess anti-inflammatory and anti-tumor activities and to be a main constituent of fennel, anise, and camphor. In the present study, we evaluated anti-metastatic and apoptotic effects of anethole on highly-metastatic HT-1080 human fibrosarcoma tumor cells. Despite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service