05292
Spermidine 0.1 M solution
Synonym(s):
Spermidine solution, Additive Screening Solution 30/Kit-No 78374
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32
Recommended Products
Quality Level
density
1.00 g/mL at 20 °C
storage temp.
2-8°C
SMILES string
NCCCCNCCCN
InChI
1S/C7H19N3/c8-4-1-2-6-10-7-3-5-9/h10H,1-9H2
InChI key
ATHGHQPFGPMSJY-UHFFFAOYSA-N
Gene Information
mouse ... Odc1(18263)
rat ... Ppm1a(24666)
Biochem/physiol Actions
Spermidine is a naturally occurring polyamine. It is associated with various cellular functions. It is responsible for the condensation and aggregation of DNA in aqueous solution. Spermidine is known to extend lifespan via epigenetic modifications, induction of cell death and inhibition of necrosis.
Spermidine 0.1M solution is an additive screening solution of Additive Screening Kit. Additive Screen kit is designed to allow rapid and convenient evaluation of additives and their ability to influence the crystallization of the sample. The Additive Kit provides a tool for refining crystallization conditions.
Spermidine 0.1M solution is an additive screening solution of Additive Screening Kit. Additive Screen kit is designed to allow rapid and convenient evaluation of additives and their ability to influence the crystallization of the sample. The Additive Kit provides a tool for refining crystallization conditions.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Induction of autophagy by spermidine promotes longevity.
Eisenberg T et al.
Nature Cell Biology, 11, 1305-1305 (2009)
Satyanarayana U
Biochemistry (2014)
Marjorie Catala et al.
Bio-protocol, 10(12), e3646-e3646 (2021-03-05)
Transfer RNAs (tRNAs) are heavily decorated with post-transcriptional modifications during their biosynthesis. To fulfil their functions within cells, tRNAs undergo a tightly controlled biogenesis process leading to the formation of mature tRNAs. In addition, functions of tRNAs are often modulated
53rd National Meeting of the Italian Society of Biochemistry and Molecular Biology (SIB) and National Meeting of Chemistry of Biological Systems ? Italian Chemical Society (SCI - Section CSB) (2008)
Stephana Carelli et al.
Experimental dermatology, 21(12), 953-955 (2012-10-24)
Hair follicle (HF) regression is characterized by the activation of apoptosis in HF cells. Dermal papilla cells play a leading role in the regulation of HF development and cycling. Human follicular dermal papilla cells (HFDPC) were used to investigate the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service