Skip to Content
Merck
All Photos(1)

Key Documents

04129

Sigma-Aldrich

1-Butyl-3-methylimidazolium chloride

≥99.0% (HPLC)

Synonym(s):

BMIMCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15ClN2
CAS Number:
Molecular Weight:
174.67
Beilstein:
6449277
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (HPLC)

form

powder

impurities

≤0.2% water

mp

~70 °C

SMILES string

[Cl-].CCCCn1cc[n+](C)c1

InChI

1S/C8H15N2.ClH/c1-3-4-5-10-7-6-9(2)8-10;/h6-8H,3-5H2,1-2H3;1H/q+1;/p-1

InChI key

FHDQNOXQSTVAIC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

1-Butyl-3-methylimidazolium chloride is a high-purity ionic liquid (HPIL) with low water content. It shows high cellulose-dissolving ability.

Application

[BMIM]Cl in combination with glycerol forms an effective plasticizer with higher thermal stability, flexibility and tensile strength.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

377.6 °F - (External MSDS)

Flash Point(C)

192 °C - (External MSDS)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal structure of 1-butyl-3-methylimidazolium chloride. A clue to the elucidation of the ionic liquid structure
Saha, Satyen, et al.
Chemistry Letters (Jpn), 32.8, 740-741 (2003)
Ionic Liquids for Catalysis
Aldrich Chemfiles, 5(6) null
Hydrolysis of cellulosic bamboo biomass into reducing sugars via a combined alkaline solution and ionic liquid pretreament steps
Kassaye, Samuel, Kamal K. Pant, and Sapna Jain
Renewable Energy, 104, 177-184 (2017)
Saccharification of cellulose in the ionic liquids and glucose recovery
Liu, Zhen, et al.
Renewable Energy (2017)
A simple method for preparation of a novel hydrophobic ionic liquid with a per-fluoro-tert-butoxide anion
Kurnia, Kiki A., et al.
New. J. Chem., 41.1, 47-50 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service