Skip to Content
Merck
All Photos(1)

Key Documents

95803

Sigma-Aldrich

1-Methyl-3-octylimidazolium chloride

≥97.0% (HPLC)

Synonym(s):

OMIMCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H23ClN2
CAS Number:
Molecular Weight:
230.78
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (HPLC)

form

liquid

refractive index

n20/D 1.505-1.515

density

1.01 g/mL at 20 °C (lit.)

SMILES string

[Cl-].CCCCCCCC[n+]1ccn(C)c1

InChI

1S/C12H23N2.ClH/c1-3-4-5-6-7-8-9-14-11-10-13(2)12-14;/h10-12H,3-9H2,1-2H3;1H/q+1;/p-1

InChI key

OXFBEEDAZHXDHB-UHFFFAOYSA-M

Application

1-Methyl-3-octylimidazolium chloride ionic liquid ([MOIM]Cl) can be used as:      
  • A reaction medium for the hydrolysis of sucrose to form hydroxymethylfurfural in the presence of HCl and metal chloride catalyst.        
  • A structure-directing agent during the preparation of organized mesoporous alumina.

Other Notes

Lipophilic ionic liquid

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aggregation and micelle formation of ionic liquids in aqueous solution
Miskolczy, Zsombor, et al.
Chemical Physics Letters, 400.4, 296-300 (2004)
Anna S Dotsenko et al.
Bioresource technology, 250, 429-438 (2017-12-02)
The pretreatment of softwood and hardwood samples (spruce and hornbeam wood) with 1-butyl-3-methylimidazolium chloride ([Bmim]Cl) was undertaken for further simultaneous enzymatic saccharification of renewable non-food lignocellulosic biomass and microbial fermentation of obtained sugars to ethanol and fumaric acid. A multienzyme
Synthesis of organized mesoporous alumina templated with ionic liquids
Zilkova N, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 95(1-3), 176-179 (2006)
Solute?solvent interactions of glycine, l-alanine, and l-valine in aqueous 1-methyl-3-octylimidazolium chloride ionic liquid solutions in the temperature interval (288.15?308.15) K
Pal, Amalendu, et al.
Thermochimica Acta, 590, 127-137 (2014)
Z.M.A. Judeh et al.
Tetrahedron Letters, 43, 9381-9381 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service