Skip to Content
Merck
All Photos(1)

Key Documents

03854

Sigma-Aldrich

Creosote from beechwood tar

Synonym(s):

Beechwood creosote

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352401
NACRES:
NA.25

Quality Level

ign. residue

≤0.01% (as SO4)

bp

200-220 °C (lit.)

transition temp

solidification point <−20 °C

density

1.09 g/mL at 20 °C

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

InChI

1S/C7H8O2/c1-9-7-5-3-2-4-6(7)8/h2-5,8H,1H3

InChI key

LHGVFZTZFXWLCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Beechwood creosote is a colorless to yellowish greasy liquid obtained from the distillation of wood tar. It comprises of phenol, 4-methylphenol, 2-methoxyphenol, and 2-methoxy-4-methylphenol as its major volatile constituents.

Application

Creosote from beechwood tar has been used as raw material in the synthesis of phenolic branched-chain fatty acids (n-PBC-FA) in the presence of a modified H+ Ferrierite zeolite catalyst.

Biochem/physiol Actions

Creosote from beechwood tar has been reported to exhibit antioxidant activity comparable to that of popular antioxidants α-tocopherol and butylated hydroxytoluene (BHT). It is used as a herbal antidiarrheal medicine owing to its antisecretory activity. Beechwood creosote has also been found to show bactericidal, fungicidal and laxative action.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1A - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Korean Beechwood Creosote as a Substitute to an Antibiotic for Post Weaning Diarrhea in Piglets.
Sodhi S, et al.,
Pakistan Veterinary Journal, 34(3) (2014)
Bio-based phenolic-branched-chain fatty acid isomers synthesized from vegetable oils and natural monophenols using modified H+-Ferrierite zeolite
Yan Z, et al.,
Industrial Crops and Products, 114 (2018)
Sari Vilhunen et al.
Journal of hazardous materials, 179(1-3), 776-782 (2010-04-13)
A re-circulated flow-through photoreactor was used to evaluate the ultraviolet (UV) photolysis and UV/H(2)O(2) oxidation process in the purification of three different water matrices. Chemically coagulated and electrocoagulated surface water, groundwater contaminated with creosote wood preservative and 1,2-dichloroethane (DCE) containing
Albert L Juhasz et al.
Environmental pollution (Barking, Essex : 1987), 158(2), 585-591 (2009-09-25)
The impact of residual PAHs (2250 +/- 71 microg total PAHs g(-1)) following enhanced natural attenuation (ENA) of creosote-contaminated soil (7767 +/- 1286 microg total PAHs g(-1)) was assessed using a variety of ecological assays. Microtox results for aqueous soil
Paolo Pironi et al.
Environmental science & technology, 45(7), 2980-2986 (2011-03-01)
Smoldering combustion has been introduced recently as a potential remediation strategy for soil contaminated by nonaqueous phase liquids (NAPLs). Published proof-of-concept experiments demonstrated that the process can be self-sustaining (i.e., requires energy input only to start the process) and achieve

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service