Skip to Content
Merck
All Photos(1)

Key Documents

00473

Sigma-Aldrich

Hydrogen

≥99.999%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2
CAS Number:
Molecular Weight:
2.02
EC Number:
MDL number:
UNSPSC Code:
12141902
PubChem Substance ID:

vapor density

0.07 (21 °C, vs air)

Quality Level

Assay

≥99.999%

autoignition temp.

1060 °F

expl. lim.

74.2 %

bp

−252.8 °C (lit.)

mp

−259.2 °C (lit.)

SMILES string

[H]

InChI

1S/H2/h1H

InChI key

UFHFLCQGNIYNRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

pressure tin; filling pressure at 15°C: 11 bar; content (15°C, 1 bar): 11 L

Other Notes

Sales restrictions may apply

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Compr. Gas

Storage Class Code

2B - Aerosol cans and lighters

WGK

nwg

Flash Point(F)

<-238.0 °F - closed cup

Flash Point(C)

< -150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick S Fier et al.
Science (New York, N.Y.), 342(6161), 956-960 (2013-11-23)
Fluorinated heterocycles are prevalent in pharmaceuticals, agrochemicals, and materials. However, reactions that incorporate fluorine into heteroarenes are limited in scope and can be hazardous. We present a broadly applicable and safe method for the site-selective fluorination of a single carbon-hydrogen
Hiroshi Ishikita et al.
Journal of the Royal Society, Interface, 11(91), 20130518-20130518 (2013-11-29)
In protein environments, proton transfer reactions occur along polar or charged residues and isolated water molecules. These species consist of H-bond networks that serve as proton transfer pathways; therefore, thorough understanding of H-bond energetics is essential when investigating proton transfer
Ondrej L Krivanek et al.
Nature, 514(7521), 209-212 (2014-10-10)
Vibrational spectroscopies using infrared radiation, Raman scattering, neutrons, low-energy electrons and inelastic electron tunnelling are powerful techniques that can analyse bonding arrangements, identify chemical compounds and probe many other important properties of materials. The spatial resolution of these spectroscopies is
Guancai Xie et al.
Advanced materials (Deerfield Beach, Fla.), 25(28), 3820-3839 (2013-07-03)
Hydrogen production from solar water splitting has been considered as an ultimate solution to the energy and environmental issues. Over the past few years, graphene has made great contribution to improving the light-driven hydrogen generation performance. This article provides a
Hou Wang et al.
Advances in colloid and interface science, 195-196, 19-40 (2013-05-07)
Graphene, as an ideal two-dimensional material and single-atom layer of graphite, has attracted exploding interests in multidisciplinary research because of its unique structure and exceptional physicochemical properties. Especially, graphene-based materials offer a wide range of potentialities for environmental remediation and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service