Skip to Content
Merck
All Photos(1)

Key Documents

RPOLSP6-RO

Roche

SP6 RNA Polymerase

from Escherichia coli BL 21/pSR3

Synonym(s):

mRNA, polymerase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

biological source

Escherichia coli (BL 21/pSR3)

Quality Level

Assay

100% (SDS-PAGE)

form

solution

specific activity

≥20 U/μL

mol wt

96  kDa (Single polypeptide chain)

packaging

pkg of 1,000 U (10810274001)
pkg of 5,000 U (11487671001)

manufacturer/tradename

Roche

technique(s)

DNA sequencing: suitable
Northern blotting: suitable
Southern blotting: suitable

color

colorless

pH

~7.9 (39 °F)

solubility

water: miscible

suitability

suitable for molecular biology

UniProt accession no.

application(s)

genomic analysis
life science and biopharma

foreign activity

Endonucleases, none detected (up to 30U using Lamda-DNA)
Endonucleases, none detected (upto 30U using MWM III-DNA )
Nicking activity, none detected (up to 30U using pBR322-DNA)
RNase, none detected (up to 30U using MS II- RNA )

storage temp.

−20°C

Gene Information

Escherichia coli ... rpoB(948488)

General description

With this system, homogeneously labeled single-stranded RNA can be generated. Transcripts can be non-radioactively labeled with biotin or DIG-11-UTP or radioactively labeled to high specific activity with [α-32P] or [α-35S] labeled nucleotides.

Contents:
  • SP6 RNA Polymerase, ≥20 U/μl, in buffer, pH 7.9
  • Transcription Buffer, 10x concentrated

Specificity

Promoter specifity

SP6 RNA polymerase is extremely promoter-specific and only transcribes bacteriophage SP6 DNA or DNA cloned downstream of a SP6 promoter (e.g., pSPTBM20; pSPTBM21).

Heat inactivation: Stop the reaction by adding 2 μl 0.2 M EDTA (pH 8.0) and/or heat to 65 °C.

Application

SP6 RNA Polymerase can transcribe RNA from cloned DNA templates that are downstream from an SP6 promoter. The synthesis can be performed with labeled NTPs to generate highly labeled RNA. Synthesized RNA can be used in many applications, including:
  • RNA or DNA blotting techniques
  • In situ hybridization
  • RNase protection studies: Transcripts synthesized by the enzyme are used as precursor RNA for studies on RNA splicing and processing.
  • Synthesis of capped RNA in vitro with addition of m7GpppG or m7GpppA in excess over GTP or ATP during the transcription reaction. The generated antisense RNA can be introduced into cells to suppress the expression of the corresponding genes.
  • The synthesis of antisense RNA probes

Quality

Test Buffer
40 mM Tris-HCl, pH 8.0 (+20°C), 6 mM MgCl2, 10 mM dithiothreitol, 2 mM spermidine, pH approximately 8.0 (+20°C).
Absence of Endonucleases
1. 1 μg lambda DNA is incubated with SP6 RNA polymerase for 4 hours at +37°C in 25 μl test buffer. The number of enzyme units which show no degradation of lambda DNA is > 30 U.
2. 1 μg Eco RI/Hind III fragments of lambda DNA is incubated with SP6 RNA polymerase for 4 hours at +37°C in 25 μl test buffer. The number of enzyme units which show no alteration of the banding pattern is > 30 U.
Absence of Nicking Activity
1 μg pBR322 DNA is incubated with SP6 RNA polymerase for 4 hours at +37°C in 25 μl test buffer. The number of enzyme units which show no relaxing of supercoiled structure is > 30.
Absence of RNases
4 μg MS2 RNA are incubated with SP6 RNA polymerase for 4 hours at +37°C in 50 μl test buffer. The number of enzyme units which show no degradation of MS2 RNA is > 30.
Performance in Transcription Assay
SP6 RNA polymerase is function tested in the SP6/T7 Transcription Kit (Cat. No. 10 999 644 001). The incorporation rate in the standard assay with 0.5 μg pST18 neo DNA linearized with Eco RI and 50 mCi [alpha-32P] CTP, [400 Ci/mmol (15 TBq/mmol)] gives >50% of the input radioactivity in 20 minutes.

Specifications

Synthesis of hybridization probes: SP6 RNA polymerase allows highly efficient production of homogeneously labeled RNA. This labeled RNA may be used as hybridization probes in Southern, northern, and dot blots, as well as in situ hybridizations.
Suitable labels: Transcripts can be nonradioactively labeled with biotin-16-UTP, DIG-11-UTP, or fluorescein-12-UTP. They may also be radioactively labeled to high specific activity with [a-32P]- or [a-35S]-labeled nucleotides.

Note: Roche has 10x concentrated RNA Labeling Mixes that are specially designed for DIG-, biotin-, and fluorescein-labeling. These mixes work well with SP6 RNA Polymerase.

Unit Definition

One unit is the enzyme activity which incorporates 1 nmol CMP in acid-precipitable RNA products within 60 minutes at +37 °C.

Volume Activity: ≥20 U/μl

Preparation Note

Activator: BSA/spermine

Storage and Stability

Keep container tightly closed in a dry and well-ventilated place

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • SP6 RNA Polymerase, in buffer, pH 7.9 ≥20 U/μl

  • Transcription Buffer 10x concentrated

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ezgi Hacisuleyman et al.
Nature structural & molecular biology, 21(2), 198-206 (2014-01-28)
RNA, including long noncoding RNA (lncRNA), is known to be an abundant and important structural component of the nuclear matrix. However, the molecular identities, functional roles and localization dynamics of lncRNAs that influence nuclear architecture remain poorly understood. Here, we
Camilo Riquelme-Guzmán et al.
eLife, 11 (2022-10-12)
Early events during axolotl limb regeneration include an immune response and the formation of a wound epithelium. These events are linked to a clearance of damaged tissue prior to blastema formation and regeneration of the missing structures. Here, we report
Adam D Langenbacher et al.
EvoDevo, 7, 9-9 (2016-04-14)
Germ cells are specified during early development and are responsible for generating gametes in the adult. After germ cells are specified, they typically migrate to a particular niche in the organism where they reside for the remainder of its lifetime.
Chunyan Li et al.
Nature communications, 12(1), 1094-1094 (2021-02-19)
Seahorses have a circum-global distribution in tropical to temperate coastal waters. Yet, seahorses show many adaptations for a sedentary, cryptic lifestyle: they require specific habitats, such as seagrass, kelp or coral reefs, lack pelvic and caudal fins, and give birth
Yongchao Liu et al.
Scientific reports, 5, 10159-10159 (2015-05-12)
Long non-coding RNAs (lncRNAs), which have evolved as important gene expression modulators, are involved in human malignancies. The down-regulation of lncRNA growth arrest specific transcript 5 (GAS5) has been reported in several cancers, however, the underlying mechanism of lncRNA GAS5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service