Skip to Content
Merck
All Photos(1)

Key Documents

10280879001

Roche

5-Bromo-2′-deoxyuridine

>97%, crystalline solid, pkg of 1 g

Synonym(s):

5-Bromo-2′-deoxyuridine, brdu, 5-BrdU, 5-Bromo-1-(2-deoxy-β-D-ribofuranosyl)uracil, 5-Bromouracil deoxyriboside, BUdR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11BrN2O5
CAS Number:
Molecular Weight:
307.10
Beilstein:
30395
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
Pricing and availability is not currently available.

Quality Level

Assay

>97%

form

crystalline solid

mol wt

307.1

packaging

pkg of 1 g

manufacturer/tradename

Roche

mp

191-194 °C (dec.) (lit.)

shipped in

wet ice

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H](C[C@@H]1O)N2C=C(Br)C(=O)NC2=O

InChI

1S/C9H11BrN2O5/c10-4-2-12(9(16)11-8(4)15)7-1-5(14)6(3-13)17-7/h2,5-7,13-14H,1,3H2,(H,11,15,16)/t5-,6+,7+/m0/s1

InChI key

WOVKYSAHUYNSMH-RRKCRQDMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

It is conventionally used as a marker for S-phase synthesis of DNA.[1][2]

Application

5-Bromo-2′-deoxyuridine has been used to label cells in the S-phase of the cell cycle[3] and primary hepatocytes during cell proliferation studies.[4]
Immunochemically detectable nucleotide that can be incorporated into DNA instead of thymidine. Used for in vivo studies of DNA synthesis[5] and for the detection of mutagenic substances, according to the sister chromatid exchange (SCE) method.

Biochem/physiol Actions

Thymidine analog used as a mutagen in genetic research. Selectively incorporated into cellular DNA during S-phase.

Quality

Purity: 97% (from N), chromatographically homogeneous

Preparation Note

Working concentration: According to literature for cell culture, a final concentration of 10 μmol/l has been used.
According to literature, 5 to 50 mg/kg body weight mice have been used for the detection of cell proliferation.

Preparation of Working Solution
Corresponding to the BrdU-labeling reagent which is included in the Roche Cell Proliferation ELISA, BrdU kits and in the Roche 5-Bromo-2′-deoxy-uridine Labeling and Detection Kits, prepare the BrdU working solution by dissolving the substance in PBS to a 10 mM stock solution (MW of BrdU = 307.1 D). For the in vivo use of BrdU dissolve in PBS; for the in vitro use of BrdU, dissolve in double-distilled water at the same 10 mM concentration.
Storage conditions (working solution): -15 to -25 °C

Other Notes

For life science research only. Not for use in diagnostic procedures.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 1B - Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yukinobu Goto et al.
American journal of physiology. Cell physiology, 285(2), C253-C259 (2003-04-04)
The present study was designed to develop methods to study the production and release of monocytes from the bone marrow using the thymidine analog 5'-bromo-2'-deoxyuridine (BrdU). Dividing monocytes in bone marrow were labeled with BrdU (MOBrdU), and their release into
Hwan-Ki Kim et al.
Experimental neurobiology, 29(6), 417-424 (2020-12-08)
The myelination of axons in the vertebrate nervous system through oligodendrocytes promotes efficient axonal conduction, which is required for the normal function of neurons. The central nervous system (CNS) can regenerate damaged myelin sheaths through the process of remyelination, but
Loss of Integrin $\alpha$v$\beta$8 in Murine Hepatocytes Accelerates Liver Regeneration
Greenhalgh SN, et al.
The American Journal of Pathology, 189(2), 258-271 (2019)
Effects of Wnt5a protein on proliferation and apoptosis in JAR choriocarcinoma cells
Peng S, et al.
Molecular Medicine Reports, 4(1), 99-104 (2011)
K M Connolly et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 41(1), 1-6 (1993-01-01)
Proliferating cell nuclear antigen (PCNA) was evaluated as a marker of cell proliferation in formalin-fixed rat liver tissue through a comparative study with the thymidine analogue 5-bromo-2'-deoxyuridine (BrdU). The comparison was conducted through the introduction of a dual immunohistochemical procedure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service