Skip to Content
Merck
All Photos(1)

Key Documents

10197734001

Roche

Glutamate Dehydrogenase (GlDH)

from beef liver

Synonym(s):

DNA marker

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204

biological source

bovine liver

Quality Level

form

lyophilized

specific activity

10 U/mg
~10 units/mg protein (at 25 °C with 2-oxoglutarate as the substrate, and ADP as the activator.)

packaging

pkg of 3,000 U

manufacturer/tradename

Roche

concentration

≥10-20 % (w/w)

technique(s)

activity assay: suitable

color

white

optimum pH

8

solubility

water: 20 mg/mL, colorless

suitability

suitable for UV spectrophotometry and general use

UniProt accession no.

application(s)

life science and biopharma

foreign activity

ADH <0.00500%
LDH <0.00500%
MDH <0.00500%

shipped in

wet ice

storage temp.

2-8°C

Gene Information

bovine ... GLUD1(281785)

Related Categories

General description

Approximately 10 U/mg lyophilizate (120 U/mg enzyme protein) at +25°C with 2-oxoglutarate as the substrate, and ADP as the activator.
GDH (Glutamate dehydrogenase) is a hexamer of 449 residues and is located in the mitochondria. It acts as a branch-point enzyme between amino acid oxidation and urea production. L-glutamate:NAD(P)+ oxidoreductase is involved in deamination.

Application

Glutamate dehydrogenase has been used to measure residual ammonium by enzymatic analysis during fermentation by wine yeast. It used as a component of cofactor recycling systems for NAD(P) and NAD(P)H.

Quality

Contaminants: <0.005% ADH, LDH, and MDH, each

Sequence

The smallest active structure of GIDH (310,000 to 350,000 ) is a hexamer subunit of 55,400 D. This hexamer reversibly self-assembles forming a polymer with up to eight hexamers (2,200 kD).

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Hughes et al.
Analytica chimica acta, 933, 82-88 (2016-08-09)
A reagentless glutamate biosensor was applied to the determination of glutamate released from liver hepatocellular carcinoma cells (HepG2) in response to toxic challenge from various concentrations of paracetamol. A screen printed carbon electrode (SPCE) containing the electrocatalyst Meldola's Blue (MB-SPCE)
Disruption of the cell wall integrity gene ECM33 results in improved fermentation by wine yeast
Jin Zhang
Metabolic engineering (2018)
T J Stillman et al.
Journal of molecular biology, 234(4), 1131-1139 (1993-12-20)
We have solved the structure of the binary complex of the glutamate dehydrogenase from Clostridium symbiosum with glutamate to 1.9 A resolution. In this complex, the glutamate side-chain lies in a pocket on the enzyme surface and a key determinant
P J O'Brien et al.
Laboratory animals, 36(3), 313-321 (2002-07-30)
In a recent study in rats, alanine aminotransferase (ALT), the preferred plasma biomarker of hepatocellular injury in rats, was ineffective at detecting marked hepatic necrosis produced by acetaminophen (Human and Experimental Toxicology 19, 277-83, 2000). In contrast, glutamate dehydrogenase (GLDH)
Po Chen et al.
Fly, 14(1-4), 3-9 (2020-07-04)
Glutamine: fructose-6-phosphate amidotransferase (GFAT) enzymes catalyse the first committed step of the hexosamine biosynthesis pathway (HBP) using glutamine and fructose-6-phosphate to form glucosamine-6-phosphate (GlcN6P). Numerous species (e.g. mouse, rat, zebrafish, chicken) including humans and Drosophila encode two broadly expressed copies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service