Skip to Content
Merck
All Photos(2)

Key Documents

QTUM000EX

Milli-Q

Quantum® Polishing Cartridge

For use with Milli-Q® Biocel / Academic / Gradient / Synthesis systems

Synonym(s):

Quantum® EX polishing cartridge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
NACRES:
JA.13

Quality Level

feature

(removes trace levels of organics and ions from ultrapure water)

packaging

pkg of 1 unit

compatibility

for use with Milli-Q® Academic
for use with Milli-Q® Biocel
for use with Milli-Q® Gradient
for use with Milli-Q® Synthesis

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

The Quantum® Pack has been designed to remove trace levels of organics and ions from ultrapure water. It is made of Organex (mix of synthetic activated carbon & virgin ion-exchange resins) and virgin ion-exchange resins.
Quantum® EX polishing cartridges can be used with Milli-Q® Biocel / Academic / Gradient / Synthesis systems.

Application

For Milli-Q® systems. Contains Organex & Ionex Resins for trace levels of organics and ions.

Features and Benefits

  • Contains Organex and Ionex resins to remove trace organic and ionic contaminants.
  • Care-free maintenance: easily installed and replaced.
  • Uses ion exchange technology for purification.
  • Should be replaced every 6 months.
  • Certificate of Quality available online.

Components

  • Organex (Mix of Synthetic activated carbon & virgin Ion exchange resins).
  • Virgin Ion Exchange Resins.

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
QUANTUM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service