Skip to Content
Merck
All Photos(1)

Key Documents

PMEF-CFL

Sigma-Aldrich

EmbryoMax® Primary Mouse Embryonic Fibroblasts

Synonym(s):

CF-1 MEF Feeder Cells, CF-1 MEFs, CF-1 PMEFs, CF1 Mouse Feeder Cells, MEF Feeder Cells, CF-1 PMEFs, CF-1 MEFs, MEFs, Mouse Feeder Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106509
eCl@ss:
32011203
NACRES:
NA.75

Pricing and availability is not currently available.

biological source

mouse

Quality Level

manufacturer/tradename

Specialty Media
EmbryoMax®

technique(s)

cell culture | stem cell: suitable

input

sample type: human embryonic stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type primary embryotic fibroblasts (PMEFs)
sample type induced pluripotent stem cell(s)

shipped in

liquid nitrogen

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
ABF32MABF276MABF854
clone

9D5.1, monoclonal

clone

polyclonal

clone

17, monoclonal

clone

6B4, monoclonal

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

biological source

mouse

biological source

rabbit

biological source

mouse

biological source

mouse

species reactivity

human, mouse

species reactivity

mouse, rat, human, horse

species reactivity

human

species reactivity

human

isotype

IgG1κ

isotype

-

isotype

IgG1κ

isotype

IgG1κ

General description

Primary Mouse Embryo Fibroblasts, Strain CF1, are not Mytomycin C treated and serve as a feeder layer for both mouse and human ES/IPS cells. The cells are provided at passage 3 and can be further expanded for about 5-6 passages and frozen for later use.

Plating MEF Feeder Cells

Procedure:

1. Prior to thawing PMEF feeder cells, coat plates/flasks with Gelatin solution.
2. Thaw PMEF vial(s) quickly in a 37 °C water bath and transfer to a 15 mL tube (already containing 10 mL of warm PMEF Feeder Cell Medium). Gently invert the tube to distribute, and centrifuge at 300 xg for 4–5 minutes.
3. Remove supernatant and resuspend the cell pellet in warm PMEF Feeder Cell Medium.
4. Remove the Gelatin solution from plates/flasks, and aliquot the PMEF feeder cell suspension at the densities recommended in Table 4.1 of the mouse ES protocol guide
5. Incubate the PMEF Feeder cells at 37 °C with 5% CO2. Use Figures 4A, B and C in the mouse ES protocol guide as a guide for an estimate of correct PMEF density and
appearance. Gelatinized plates may be used for 12–14 days.
The EmbryoMax range of PMEF cells provides researchers with a convenient solution for ES cell culture by eliminating the need for time consuming feeder cell isolation and preparation. Many embryonic stem cell culture protocols necessitate the use of primary mouse embryo fibroblast (PMEF) cells. In these protocols, ES cells are typically cultured on a monolayer of PMEF feeder cells. Feeder cells perform two important roles in stem cell culture: they secrete several important growth factors into the medium, which help maintain pluripotency, and they provide a cellular matrix for ES cells to grow.

Cell Line Description

Primary Mouse Embryo Fibroblasts

Packaging

5-6x106 ea

Physical form

The cells are provided frozen into individual vials, containing approximately 5-6 x 106 fibroblasts per vial, and are sold as a pack of 5 vials and supplied at passage 3. See ES Stem Cell Manual for complete culture and medias for PMEF cells.

Storage and Stability

After receipt, vials should be stored at -80°C. If storing for longer than 6 months, store in the vapor phase of liquid nitrogen (make sure the cap is tight) for up to 2 years.

Legal Information

EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Customers Also Viewed

    Articles

    Mouse embryonic fibroblasts (MEFs) serve as a feeder layer for both mouse and human embryonic stem cells (ES cells) and induced pluripotent stem cells (iPSCs).

    Mouse embryonic fibroblasts (MEFs) serve as a feeder layer for both mouse and human embryonic stem cells (ES cells) and induced pluripotent stem cells (iPSCs).

    Mouse embryonic fibroblasts (MEFs) serve as a feeder layer for both mouse and human embryonic stem cells (ES cells) and induced pluripotent stem cells (iPSCs).

    Mouse embryonic fibroblasts (MEFs) serve as a feeder layer for both mouse and human embryonic stem cells (ES cells) and induced pluripotent stem cells (iPSCs).

    Protocols

    Stem Cell protocols for cryopreservation, thawing of cryopreserved stem cells and media preparation.

    This technical article covers the indirect co-culture of embryonic stem cells with embryonic fibroblasts.

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service