Skip to Content
Merck
All Photos(4)

Key Documents

PICM03050

Millipore

Millicell® Standing Cell Culture Inserts

pore size 0.4 μm, diam. 30 mm, transparent PTFE membrane, hydrophilic, H 13 mm, size 6 wells, sterile

Synonym(s):

Millicell Cell Culture Insert, 30 mm, hydrophilic PTFE, 0.4 µm, Millicell-CM, cell culture inserts, permeable culture insert, plate inserts, tissue culture insert, tissue culture plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32011202
NACRES:
NB.22

material

polystyrene housing
transparent PTFE membrane

Quality Level

sterility

ethylene oxide treated
sterile

feature

hydrophilic

packaging

pack of 50

manufacturer/tradename

Millicell®

parameter

50 °C max. temp.

technique(s)

cell attachment: suitable
cell culture | mammalian: suitable
cell differentiation: suitable

H

13 mm

diam.

30 mm

filtration area

4.2 cm2

size

6 wells

surface area

4.2 cm2

working volume

1.5 mL

color

transparent, when wetted

matrix

Biopore

pore size

0.4 μm

binding type

low binding surface

detection method

fluorometric

shipped in

ambient

General description

Device Configuration: 6-well plate, 0.4μm pore size

With Millicell® inserts, attachment or suspension cells can access media from both their apical and basolateral sides. Cell growth, structure, and function more closely mimic what occurs in vivo. In addition, Millicell® inserts make it possible to study both sides of the cell monolayer.

Millicell® Standing Inserts:
- Promotes excellent cell growth and provides an exceptional opportunity for cell studies

Membrane Type:
Biopore Membrane (hydrophilic PTFE)
- For low protein-binding, live cell viewing, and immunofluorescent applications

Applications
Cell Attachment, Cell Growth, Cell Differentiation , Immunocytochemistry

Application

Hydrophilic PTFE cell culture insert with pore size of 0.4 µm used in a 6-well plate for Cell Attachment, Cell Culture, Cell Differentiation, Fluorescence & Low Protein Binding.
Research Category
Cell Culture

Packaging

Individually blister packaged

Legal Information

BIOPORE is a trademark of Merck KGaA, Darmstadt, Germany
Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CB1 receptors and post-ischemic brain damage: Studies on the toxic and neuroprotective effects of cannabinoids in rat organotypic hippocampal slices.
Landucci, Elisa, et al.
Neuropharmacology, 60, 674-682 (2011)
Synapse elimination in olivo-cerebellar explants occurs during a critical period and leaves an indelible trace in Purkinje cells.
Letellier, Mathieu, et al.
Proceedings of the National Academy of Sciences of the USA, 106, 14102-14107 (2009)
Daniela Buonvicino et al.
British journal of pharmacology, 177(14), 3342-3356 (2020-03-22)
Drugs able to counteract progressive multiple sclerosis (MS) represent a largely unmet therapeutic need. Even though the pathogenesis of disease evolution is still obscure, accumulating evidence indicates that mitochondrial dysfunction plays a causative role in neurodegeneration and axonopathy in progressive
Preparation of organotypic hippocampal slice cultures: interface method.
De Simoni, Anna and Yu, Lily M Y
Nature Protocols, 1, 1439-1445 (2006)
R Le Bouffant et al.
Human reproduction (Oxford, England), 25(10), 2579-2590 (2010-07-31)
The initiation of meiosis is crucial to fertility. While extensive studies in rodents have enhanced our understanding of this process, studies in human fetal ovary are lacking. We used RT-PCR and immunohistochemistry to investigate expression of meiotic factors in human

Protocols

This protocol covers three methods for the microscopic examination of cell samples.

This technical article covers the indirect co-culture of embryonic stem cells with embryonic fibroblasts.

This page covers the basic indirect co-culture procedure on both sides of Millicell cell culture insert membranes.

This page covers the basic indirect co-culture procedure on both sides of Millicell cell culture insert membranes.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service