PC234L
Anti-Vasopressin Rabbit pAb
lyophilized, Calbiochem®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.43
Recommended Products
biological source
rabbit
Quality Level
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
form
lyophilized
contains
≤0.1% sodium azide as preservative
species reactivity
rat
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
isotype
IgG
shipped in
ambient
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
rat ... Avp(24221)
General description
Rabbit polyclonal antibody supplied as lyophilized undiluted serum. Recognizes vasopressin.
Recognizes vasopressin in rat hypothalamus.
This Anti-Vasopressin Rabbit pAb is validated for use in Frozen Sections, Immunofluorescence for the detection of Vasopressin.
Immunogen
Rat
rat arginine vasopressin
Application
![](/deepweb/assets/sigmaaldrich/marketing/global/images/pdp-descriptions/emd-biosciences-images-stain-en-us-pc234l-stain/emd-biosciences-images-stain-en-us-pc234l-stain.jpg)
Frozen Sections (1:400-1:800, Cy3; 1:2000-1:4000, Bn-SA/HRP)
Immunofluorescence (1:200-1:400)
Warning
Toxicity: Standard Handling (A)
Physical form
Undiluted serum.
Reconstitution
Reconstitute the lyophilized antibody with 100 µl sterile distilled H₂O. Be careful to reconstitute the entire contents of the vial; during shipment and handling portions of the lyophilized pellet may have become dislodged and may not be in the bottom of the vial. Dilute with sterile PBS or Tris buffer at dilutions no higher than 1:10. Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months. Avoid freeze/thaw cycles of solutions.
Analysis Note
Positive Control
Rat hypothalamus
Rat hypothalamus
Other Notes
Antibody specificity was examined in the rat paraventricular nucleus and supraoptic nucleus. Staining is completely eliminated by pretreatment of antibody with arginine vasopressin (10 µg/ml). Preabsorption of antibody with oxytocin (100 µg/ml) resulted in no loss of immunostaining. Antibody should be titrated for optimal results in individual systems.
Sudhof, T.C. 1995. Nature375, 645.
Hokfelt, T., et al. 1987. Experientia Suppl.56, 154.
Hokfelt, T., et al. 1987. Experientia Suppl.56, 154.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yarely C Hoffiz et al.
Scientific reports, 11(1), 2335-2335 (2021-01-29)
Long-standing clinical findings report a dramatic surge of vasopressin in umbilical cord blood of the human neonate, but the neural underpinnings and function(s) of this phenomenon remain obscure. We studied neural activation in perinatal mice and rats, and found that
V Tobin et al.
Journal of neuroendocrinology, 24(4), 629-641 (2011-10-13)
In magnocellular neurones of the supraoptic nucleus (SON), the neuropeptides vasopressin and oxytocin are synthesised and packaged into large dense-cored vesicles (LDCVs). These vesicles undergo regulated exocytosis from nerve terminals in the posterior pituitary gland and from somata/dendrites in the
Zita Liutkevičiūtė et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, fj201800443-fj201800443 (2018-06-26)
Ants are emerging model systems to study cellular signaling because distinct castes possess different physiologic phenotypes within the same colony. Here we studied the functionality of inotocin signaling, an insect ortholog of mammalian oxytocin (OT), which was recently discovered in
Leqiang Sun et al.
Molecular neurodegeneration, 14(1), 8-8 (2019-02-10)
Neurotropic virus-based tracers have been extensively applied in mapping and manipulation of neural circuits. However, their neurotropic and neurotoxic properties remain to be fully characterized. Through neural circuit tracing, we systematically compared the neurotropism discrepancy among different multi-trans-synaptic and mono-synaptic
Douglas W Wacker et al.
The Journal of physiology, 588(Pt 23), 4705-4717 (2010-10-06)
The anterior olfactory nucleus (AON), a component of the main olfactory system, is a cortical region that processes olfactory information and acts as a relay between the main olfactory bulbs and higher brain regions such as the piriform cortex. Utilizing
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service