Skip to Content
Merck
All Photos(6)

Key Documents

MAB318

Sigma-Aldrich

Anti-Tyrosine Hydroxylase Antibody, clone LNC1

ascites fluid, clone LNC1, Chemicon®

Synonym(s):

TH, Tyrosine Monooxygenase, Tyrosine 3-monooxygenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

LNC1, monoclonal

species reactivity

zebrafish, mouse, rat, vole, human, frog, lizard, monkey, chicken

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TH(7054)
rat ... Th(25085)

General description

Tyrosine hydroxylase plays an important role in the physiology of adrenergic neurons. It is the first and rate-limiting enzyme involved in the biosynthesis of the catecholamines Dopamine and Norepinephrine from tyrosine. TH is, therefore, a useful marker for dopaminergic and noradrenergic neurons. The enzymatic activity of TH requires ferrous ions as cofactors and is believed to be regulated by phosphorylation. At least four isoforms of human TH have been identified which result from alternative splicing.

Specificity

Recognizes an epitope on the outside of the regulatory N-terminus. Recognizes a protein of approximately 59-61 kDa by Western blot. Does not react with the following on Western Blots: dopamine-beta-hydroxylase, phenylalanine hydroxylase, trytophan hydroxylase, dehydropteridine reductase, sepiapterin reductase or phenethanolamine-N-methyl transferase (PNMT).

Immunogen

Tyrosine Hydroxylase purified from PC12 cells

Application

Detect Tyrosine Hydroxylase using this Anti-Tyrosine Hydroxylase Antibody, clone LNC1 validated for use in IH, IH(P), IP & WB with more than 85 product citations.
Immunohistochemistry(paraffin):
A 1:200-1:400 dilution of a previous lot was used in IH. 4% PFA fixed, frozen sections; 4% PFA fixed, paraffin sections 1:100 (Barrachina, M. et al., 2003). For paraffin sections Barrachina reported successful staining with microwave citrate acid antigen recovery, however other methods can likely be used as well.

Immunoprecipitation:
A previous lot of this antibody was used in IP (Perez, 2002).

Optimal working dilutions and protocols must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neuronal & Glial Markers

Quality

Routinely evaluated by Western Blot on Mouse Brain lysates.

Western blot:
1:1000 dilution of this lot detected Tyrosine Hydroxylase on 10 μg of Mouse Brain lysates.

Target description

59-63 kDa

Physical form

Ascites mouse monoclonal IgG1κ fluid containing 3% BSA. Contains no preservative.
Unpurified

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.

Analysis Note

Control
Human brain tissue, extracts from 3T3 cells treated with Forskolin (40 nM, 30 min).

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D W Smith et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 1(5 Pt 1), 486-491 (2000-08-10)
Lesch-Nyhan disease (LND) is an X-linked metabolic disorder caused by lack of activity of the purine salvage enzyme hypoxanthine phosphoribosyltransferase (HPRT) and characterized by hyperuricemia and debilitating neurological manifestations. The mechanisms underlying the neuropathology are not well understood and the
Natalia Omelchenko et al.
The Journal of comparative neurology, 494(6), 863-875 (2005-12-31)
Cholinergic afferents to the ventral tegmental area (VTA) contribute substantially to the regulation of motivated behaviors and the rewarding properties of nicotine. These actions are believed to involve connections with dopamine (DA) neurons projecting to the nucleus accumbens (NAc). However
Erika Allen et al.
Neuroscience letters, 489(1), 1-4 (2010-11-30)
We wished to determine whether L-DOPA, a common treatment for the motor deficits in Parkinson's disease, could also reverse the motor deficits that occur during aging. We assessed motor performance in young (2-3 months) and old (20-21 months) male C57BL/6
Judith Joyce Balcita-Pedicino et al.
The Journal of comparative neurology, 519(6), 1143-1164 (2011-02-24)
The lateral habenula (LHb) provides an important source of negative reinforcement signals to midbrain dopamine (DA) cells in the substantia nigra and ventral tegmental area (VTA). This profound and consistent inhibitory influence involves a disynaptic connection from glutamate neurons in
Genrieta Bochorishvili et al.
The Journal of comparative neurology, 520(5), 1047-1061 (2011-09-22)
The retrotrapezoid nucleus (RTN) contains CO(2) -responsive neurons that regulate breathing frequency and amplitude. These neurons (RTN-Phox2b neurons) contain the transcription factor Phox2b, vesicular glutamate transporter 2 (VGLUT2) mRNA, and a subset contains preprogalanin mRNA. We wished to determine whether

Articles

Explore the basics of working with antibodies including technical information on structure, classes, and normal immunoglobulin ranges.

Explore the basics of working with antibodies including technical information on structure, classes, and normal immunoglobulin ranges.

Antibodies combine with specific antigens to generate an exclusive antibody-antigen complex. Learn about the nature of this bond and its use as a molecular tag for research.

Antibodies combine with specific antigens to generate an exclusive antibody-antigen complex. Learn about the nature of this bond and its use as a molecular tag for research.

See All

Protocols

Tips and troubleshooting for FFPE and frozen tissue immunohistochemistry (IHC) protocols using both brightfield analysis of chromogenic detection and fluorescent microscopy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service